Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 81483000-81483263 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FABP4
Entrez Gene ID
2167
Gene Name
fatty acid binding protein 4
Gene Aliases
A-FABP, AFABP, ALBP, HEL-S-104, aP2
UniGene
Hs.391561
Gene Chromosome Location
Chr.8: 81478497-81483238 on Build GRCh38
Transcripts
NM_001442.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3665339 c.73+2T>G p.? Chr.8: 81483093-81483093 on Build GRCh38
1205971 c.49T>G p.F17V Chr.8: 81483119-81483119 on Build GRCh38
6908151 c.20G>C p.G7A Chr.8: 81483148-81483148 on Build GRCh38
4975946 c.7G>A p.D3N Chr.8: 81483161-81483161 on Build GRCh38
4840978 c.1A>G p.M1V Chr.8: 81483167-81483167 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs551187942 Chr.8: 81483046-81483046 on Build GRCh38
rs372425566 Chr.8: 81483048-81483048 on Build GRCh38
rs979113075 Chr.8: 81483082-81483082 on Build GRCh38
rs113680038 Chr.8: 81483236-81483236 on Build GRCh38
rs773445778 Chr.8: 81483093-81483093 on Build GRCh38
rs375524818 Chr.8: 81483171-81483171 on Build GRCh38
rs112251841 Chr.8: 81483235-81483235 on Build GRCh38
rs959655703 Chr.8: 81483223-81483223 on Build GRCh38
rs775686685 Chr.8: 81483180-81483180 on Build GRCh38
rs778995342 Chr.8: 81483051-81483051 on Build GRCh38
rs757126990 Chr.8: 81483215-81483215 on Build GRCh38
rs749658927 Chr.8: 81483118-81483118 on Build GRCh38
rs753616843 Chr.8: 81483212-81483212 on Build GRCh38
rs746621810 Chr.8: 81483083-81483083 on Build GRCh38
rs745316056 Chr.8: 81483055-81483055 on Build GRCh38
rs111976109 Chr.8: 81483234-81483234 on Build GRCh38
rs760792992 Chr.8: 81483200-81483200 on Build GRCh38
rs1031282925 Chr.8: 81483041-81483041 on Build GRCh38
rs771371189 Chr.8: 81483163-81483163 on Build GRCh38
rs757881560 Chr.8: 81483068-81483068 on Build GRCh38
rs764440778 Chr.8: 81483209-81483209 on Build GRCh38
rs926908618 Chr.8: 81483222-81483222 on Build GRCh38
rs767463142 Chr.8: 81483178-81483178 on Build GRCh38
rs146656915 Chr.8: 81483199-81483199 on Build GRCh38

View Full Product Details