Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6697571-6697834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIANP
Entrez Gene ID
196500
Gene Name
PILR alpha associated neural protein
Gene Aliases
C12orf53, LEDA1, PANP, leda-1
UniGene
Hs.44067
Gene Chromosome Location
Chr.12: 6689726-6700843 on Build GRCh38
Transcripts
NM_001244014.1 NM_001244015.1 NM_153685.3 XM_011520926.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
468821 c.159G>A p.S53S Chr.12: 6697651-6697651 on Build GRCh38
4044404 c.199C>T p.P67S Chr.12: 6697611-6697611 on Build GRCh38
5662149 c.104C>T p.S35F Chr.12: 6697706-6697706 on Build GRCh38
5841774 c.43C>T p.L15F Chr.12: 6697767-6697767 on Build GRCh38
4044405 c.33G>A p.L11L Chr.12: 6697777-6697777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753413912 Chr.12: 6697735-6697735 on Build GRCh38
rs777361114 Chr.12: 6697636-6697636 on Build GRCh38
rs762749279 Chr.12: 6697698-6697698 on Build GRCh38
rs573646223 Chr.12: 6697596-6697596 on Build GRCh38
rs1001249179 Chr.12: 6697715-6697715 on Build GRCh38
rs751583768 Chr.12: 6697709-6697709 on Build GRCh38
rs945813372 Chr.12: 6697652-6697652 on Build GRCh38
rs755798138 Chr.12: 6697731-6697731 on Build GRCh38
rs781539439 Chr.12: 6697787-6697787 on Build GRCh38
rs766047153 Chr.12: 6697595-6697595 on Build GRCh38
rs185182940 Chr.12: 6697672-6697672 on Build GRCh38
rs748279103 Chr.12: 6697802-6697802 on Build GRCh38
rs754866273 Chr.12: 6697604-6697604 on Build GRCh38
rs938769800 Chr.12: 6697623-6697623 on Build GRCh38
rs924673080 Chr.12: 6697601-6697601 on Build GRCh38
rs780957555 Chr.12: 6697611-6697611 on Build GRCh38
rs769049047 Chr.12: 6697673-6697673 on Build GRCh38
rs777111515 Chr.12: 6697733-6697733 on Build GRCh38
rs773370587 Chr.12: 6697696-6697696 on Build GRCh38
rs756767839 Chr.12: 6697741-6697741 on Build GRCh38
rs35524740 Chr.12: 6697714-6697715 on Build GRCh38
rs190504635 Chr.12: 6697675-6697675 on Build GRCh38
rs564469898 Chr.12: 6697729-6697729 on Build GRCh38
rs752460451 Chr.12: 6697632-6697632 on Build GRCh38
rs375366555 Chr.12: 6697812-6697812 on Build GRCh38
rs947923600 Chr.12: 6697666-6697666 on Build GRCh38
rs762107186 Chr.12: 6697686-6697686 on Build GRCh38
rs780349942 Chr.12: 6697651-6697651 on Build GRCh38
rs540288007 Chr.12: 6697811-6697811 on Build GRCh38
rs759397397 Chr.12: 6697711-6697711 on Build GRCh38
rs533469975 Chr.12: 6697808-6697808 on Build GRCh38
rs766393442 Chr.12: 6697700-6697700 on Build GRCh38
rs896317961 Chr.12: 6697644-6697644 on Build GRCh38
rs867345037 Chr.12: 6697720-6697720 on Build GRCh38
rs144462126 Chr.12: 6697633-6697633 on Build GRCh38
rs769696247 Chr.12: 6697694-6697694 on Build GRCh38
rs747583773 Chr.12: 6697768-6697768 on Build GRCh38
rs779379406 Chr.12: 6697797-6697799 on Build GRCh38
rs780507626 Chr.12: 6697767-6697767 on Build GRCh38
rs371402346 Chr.12: 6697786-6697786 on Build GRCh38
rs772668070 Chr.12: 6697649-6697649 on Build GRCh38
rs931207482 Chr.12: 6697618-6697618 on Build GRCh38
rs556398822 Chr.12: 6697642-6697642 on Build GRCh38
rs767474631 Chr.12: 6697717-6697717 on Build GRCh38

View Full Product Details