Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13955729-13955974 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF15
Entrez Gene ID
90379
Gene Name
DDB1 and CUL4 associated factor 15
Gene Aliases
C19orf72
UniGene
Hs.443636
Gene Chromosome Location
Chr.19: 13952507-13961444 on Build GRCh38
Transcripts
NM_138353.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6640460 c.424G>A p.V142I Chr.19: 13955969-13955969 on Build GRCh38
7200273 c.407G>A p.S136N Chr.19: 13955952-13955952 on Build GRCh38
4490828 c.369C>T p.V123V Chr.19: 13955914-13955914 on Build GRCh38
4882167 c.373C>A p.Q125K Chr.19: 13955918-13955918 on Build GRCh38
7164027 c.392A>G p.D131G Chr.19: 13955937-13955937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs974853972 Chr.19: 13955790-13955790 on Build GRCh38
rs748201960 Chr.19: 13955884-13955884 on Build GRCh38
rs942957037 Chr.19: 13955789-13955789 on Build GRCh38
rs767060881 Chr.19: 13955922-13955922 on Build GRCh38
rs749174376 Chr.19: 13955868-13955868 on Build GRCh38
rs1051693731 Chr.19: 13955792-13955792 on Build GRCh38
rs575445169 Chr.19: 13955815-13955815 on Build GRCh38
rs906447625 Chr.19: 13955805-13955805 on Build GRCh38
rs929184505 Chr.19: 13955759-13955759 on Build GRCh38
rs564462182 Chr.19: 13955836-13955836 on Build GRCh38
rs745540420 Chr.19: 13955864-13955864 on Build GRCh38
rs958205555 Chr.19: 13955950-13955950 on Build GRCh38
rs772304767 Chr.19: 13955885-13955885 on Build GRCh38
rs941809282 Chr.19: 13955858-13955858 on Build GRCh38
rs771069489 Chr.19: 13955892-13955892 on Build GRCh38
rs372063043 Chr.19: 13955869-13955869 on Build GRCh38
rs73515570 Chr.19: 13955865-13955865 on Build GRCh38
rs773547316 Chr.19: 13955892-13955892 on Build GRCh38
rs1017472235 Chr.19: 13955847-13955847 on Build GRCh38
rs755858495 Chr.19: 13955929-13955929 on Build GRCh38
rs1046197799 Chr.19: 13955794-13955794 on Build GRCh38
rs375325527 Chr.19: 13955938-13955938 on Build GRCh38
rs893870092 Chr.19: 13955851-13955851 on Build GRCh38
rs1047491103 Chr.19: 13955848-13955848 on Build GRCh38
rs777112520 Chr.19: 13955906-13955906 on Build GRCh38
rs371394365 Chr.19: 13955909-13955909 on Build GRCh38
rs750044694 Chr.19: 13955925-13955925 on Build GRCh38
rs546043544 Chr.19: 13955825-13955825 on Build GRCh38
rs749048995 Chr.19: 13955947-13955947 on Build GRCh38
rs1007804166 Chr.19: 13955821-13955821 on Build GRCh38
rs369087143 Chr.19: 13955901-13955901 on Build GRCh38
rs73515571 Chr.19: 13955867-13955867 on Build GRCh38
rs200397561 Chr.19: 13955953-13955953 on Build GRCh38
rs879745978 Chr.19: 13955857-13955857 on Build GRCh38
rs756705941 Chr.19: 13955921-13955921 on Build GRCh38
rs375356197 Chr.19: 13955900-13955900 on Build GRCh38
rs774163205 Chr.19: 13955874-13955874 on Build GRCh38
rs753232454 Chr.19: 13955916-13955916 on Build GRCh38
rs749224926 Chr.19: 13955907-13955907 on Build GRCh38
rs922943557 Chr.19: 13955791-13955791 on Build GRCh38

View Full Product Details