Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 55822307-55822580 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC14
Entrez Gene ID
85406
Gene Name
DnaJ heat shock protein family (Hsp40) member C14
Gene Aliases
DNAJ, DRIP78, HDJ3, LIP6
UniGene
Hs.709320
Gene Chromosome Location
Chr.12: 55820960-55829636 on Build GRCh38
Transcripts
NM_032364.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7231758 c.1814G>T p.R605L Chr.12: 55822457-55822457 on Build GRCh38
1250448 c.1870C>T p.R624W Chr.12: 55822401-55822401 on Build GRCh38
3704263 c.1800G>C p.W600C Chr.12: 55822471-55822471 on Build GRCh38
5007050 c.1859C>A p.S620* Chr.12: 55822412-55822412 on Build GRCh38
4043379 c.1803T>C p.A601A Chr.12: 55822468-55822468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778181380 Chr.12: 55822451-55822451 on Build GRCh38
rs901707755 Chr.12: 55822426-55822426 on Build GRCh38
rs376176345 Chr.12: 55822379-55822379 on Build GRCh38
rs761658646 Chr.12: 55822519-55822519 on Build GRCh38
rs752620929 Chr.12: 55822351-55822351 on Build GRCh38
rs765608994 Chr.12: 55822492-55822495 on Build GRCh38
rs1022516928 Chr.12: 55822486-55822486 on Build GRCh38
rs1040534268 Chr.12: 55822347-55822347 on Build GRCh38
rs370082867 Chr.12: 55822538-55822538 on Build GRCh38
rs763983079 Chr.12: 55822512-55822512 on Build GRCh38
rs758193397 Chr.12: 55822474-55822474 on Build GRCh38
rs572333357 Chr.12: 55822369-55822369 on Build GRCh38
rs377717282 Chr.12: 55822515-55822515 on Build GRCh38
rs747373955 Chr.12: 55822458-55822458 on Build GRCh38
rs997335973 Chr.12: 55822443-55822443 on Build GRCh38
rs781027358 Chr.12: 55822380-55822380 on Build GRCh38
rs772517438 Chr.12: 55822442-55822442 on Build GRCh38
rs750129470 Chr.12: 55822399-55822399 on Build GRCh38
rs763945072 Chr.12: 55822364-55822364 on Build GRCh38
rs759768774 Chr.12: 55822500-55822500 on Build GRCh38
rs748686971 Chr.12: 55822425-55822425 on Build GRCh38
rs369515981 Chr.12: 55822491-55822491 on Build GRCh38
rs960807018 Chr.12: 55822533-55822533 on Build GRCh38
rs758271799 Chr.12: 55822353-55822353 on Build GRCh38
rs374231851 Chr.12: 55822505-55822505 on Build GRCh38
rs180777635 Chr.12: 55822390-55822390 on Build GRCh38
rs757492193 Chr.12: 55822477-55822478 on Build GRCh38
rs776995227 Chr.12: 55822524-55822524 on Build GRCh38
rs1026656833 Chr.12: 55822454-55822454 on Build GRCh38
rs759574447 Chr.12: 55822342-55822342 on Build GRCh38
rs755633581 Chr.12: 55822401-55822401 on Build GRCh38
rs776787427 Chr.12: 55822494-55822494 on Build GRCh38
rs35436485 Chr.12: 55822441-55822442 on Build GRCh38
rs776660972 Chr.12: 55822334-55822334 on Build GRCh38
rs61758445 Chr.12: 55822457-55822457 on Build GRCh38
rs755899802 Chr.12: 55822544-55822544 on Build GRCh38
rs185940197 Chr.12: 55822542-55822542 on Build GRCh38
rs779718523 Chr.12: 55822423-55822423 on Build GRCh38
rs775497307 Chr.12: 55822506-55822506 on Build GRCh38
rs762933417 Chr.12: 55822509-55822509 on Build GRCh38
rs765325523 Chr.12: 55822348-55822348 on Build GRCh38

View Full Product Details