Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 73017231-73017460 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCHO2
Entrez Gene ID
115548
Gene Name
FCH domain only 2
Gene Aliases
-
UniGene
Hs.165762
Gene Chromosome Location
Chr.5: 72955981-73090522 on Build GRCh38
Transcripts
NM_001146032.1 NM_138782.2 XM_017009016.1 XM_017009017.1 XM_017009018.1 XM_017009019.1 XM_017009020.1 XM_017009021.1 XM_017009022.1 XM_017009023.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5512114 c.729T>C p.A243A Chr.5: 73017241-73017241 on Build GRCh38
6171531 c.726G>C p.M242I Chr.5: 73017238-73017238 on Build GRCh38
6171533 c.726G>C p.M242I Chr.5: 73017238-73017238 on Build GRCh38
1438496 c.758A>C p.K253T Chr.5: 73017270-73017270 on Build GRCh38
3856221 c.747T>C p.S249S Chr.5: 73017259-73017259 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759685920 Chr.5: 73017348-73017348 on Build GRCh38
rs190902848 Chr.5: 73017355-73017355 on Build GRCh38
rs778647518 Chr.5: 73017288-73017288 on Build GRCh38
rs944666262 Chr.5: 73017318-73017318 on Build GRCh38
rs936103617 Chr.5: 73017345-73017345 on Build GRCh38
rs529110508 Chr.5: 73017298-73017298 on Build GRCh38
rs775786888 Chr.5: 73017351-73017351 on Build GRCh38
rs527571458 Chr.5: 73017398-73017398 on Build GRCh38
rs893667603 Chr.5: 73017396-73017396 on Build GRCh38
rs1054019187 Chr.5: 73017354-73017354 on Build GRCh38
rs868415893 Chr.5: 73017276-73017276 on Build GRCh38
rs1042261721 Chr.5: 73017422-73017422 on Build GRCh38
rs778102645 Chr.5: 73017321-73017321 on Build GRCh38
rs771400058 Chr.5: 73017340-73017340 on Build GRCh38
rs765361267 Chr.5: 73017380-73017380 on Build GRCh38
rs781357611 Chr.5: 73017411-73017411 on Build GRCh38
rs945276473 Chr.5: 73017404-73017404 on Build GRCh38
rs892702455 Chr.5: 73017391-73017391 on Build GRCh38
rs776964534 Chr.5: 73017342-73017342 on Build GRCh38
rs772427448 Chr.5: 73017316-73017316 on Build GRCh38
rs747567278 Chr.5: 73017339-73017339 on Build GRCh38
rs769706330 Chr.5: 73017349-73017349 on Build GRCh38
rs777327288 Chr.5: 73017333-73017334 on Build GRCh38
rs754403480 Chr.5: 73017265-73017265 on Build GRCh38
rs547212445 Chr.5: 73017353-73017353 on Build GRCh38

View Full Product Details