Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111911377-111911638 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRYAB
Entrez Gene ID
1410
Gene Name
crystallin alpha B
Gene Aliases
CMD1II, CRYA2, CTPP2, CTRCT16, HEL-S-101, HSPB5, MFM2
UniGene
Hs.53454
Gene Chromosome Location
Chr.11: 111908620-111913213 on Build GRCh38
Transcripts
NM_001289807.1 NM_001289808.1 NM_001885.2 XM_011542608.1 XM_011542609.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139929 c.137C>T p.P46L Chr.11: 111911588-111911588 on Build GRCh38
6635830 c.133A>T p.S45C Chr.11: 111911592-111911592 on Build GRCh38
4017835 c.102G>A p.E34E Chr.11: 111911623-111911623 on Build GRCh38
6635831 c.149G>A p.R50Q Chr.11: 111911576-111911576 on Build GRCh38
396840 c.195C>G p.L65L Chr.11: 111911530-111911530 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993133728 Chr.11: 111911447-111911447 on Build GRCh38
rs782447525 Chr.11: 111911500-111911500 on Build GRCh38
rs368077774 Chr.11: 111911494-111911494 on Build GRCh38
rs782122417 Chr.11: 111911606-111911606 on Build GRCh38
rs2228387 Chr.11: 111911560-111911560 on Build GRCh38
rs781882384 Chr.11: 111911599-111911599 on Build GRCh38
rs782293258 Chr.11: 111911506-111911506 on Build GRCh38
rs782356793 Chr.11: 111911537-111911537 on Build GRCh38
rs1035012668 Chr.11: 111911419-111911419 on Build GRCh38
rs782054136 Chr.11: 111911593-111911593 on Build GRCh38
rs782151852 Chr.11: 111911551-111911551 on Build GRCh38
rs781986102 Chr.11: 111911558-111911558 on Build GRCh38
rs782366958 Chr.11: 111911507-111911507 on Build GRCh38
rs782481381 Chr.11: 111911608-111911608 on Build GRCh38
rs387907338 Chr.11: 111911559-111911559 on Build GRCh38
rs913123527 Chr.11: 111911450-111911450 on Build GRCh38
rs781846534 Chr.11: 111911576-111911576 on Build GRCh38
rs2234704 Chr.11: 111911573-111911573 on Build GRCh38
rs2234703 Chr.11: 111911603-111911603 on Build GRCh38
rs960010330 Chr.11: 111911421-111911421 on Build GRCh38
rs374981255 Chr.11: 111911605-111911605 on Build GRCh38
rs931730154 Chr.11: 111911591-111911591 on Build GRCh38
rs782489685 Chr.11: 111911475-111911475 on Build GRCh38
rs143101974 Chr.11: 111911527-111911527 on Build GRCh38
rs375886627 Chr.11: 111911538-111911538 on Build GRCh38
rs372923942 Chr.11: 111911501-111911501 on Build GRCh38
rs782720245 Chr.11: 111911595-111911595 on Build GRCh38
rs782207762 Chr.11: 111911510-111911510 on Build GRCh38
rs145768025 Chr.11: 111911610-111911610 on Build GRCh38
rs117865217 Chr.11: 111911415-111911415 on Build GRCh38
rs782547574 Chr.11: 111911601-111911601 on Build GRCh38
rs149787233 Chr.11: 111911609-111911609 on Build GRCh38
rs781935850 Chr.11: 111911543-111911543 on Build GRCh38
rs782423443 Chr.11: 111911553-111911553 on Build GRCh38

View Full Product Details