Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 20135647-20135920 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1AX
Entrez Gene ID
1964
Gene Name
eukaryotic translation initiation factor 1A, X-linked
Gene Aliases
EIF1A, EIF1AP1, EIF4C, eIF-1A, eIF-4C
UniGene
Hs.522590
Gene Chromosome Location
Chr.X: 20124518-20141848 on Build GRCh38
Transcripts
NM_001412.3

SNP Detail

SNP ID Chromosome Location
rs749404893 Chr.X: 20135718-20135718 on Build GRCh38
rs746456856 Chr.X: 20135868-20135868 on Build GRCh38
rs769320235 Chr.X: 20135885-20135886 on Build GRCh38
rs771092459 Chr.X: 20135865-20135865 on Build GRCh38
rs373498796 Chr.X: 20135724-20135724 on Build GRCh38
rs775154053 Chr.X: 20135693-20135693 on Build GRCh38
rs750420398 Chr.X: 20135850-20135850 on Build GRCh38
rs772574260 Chr.X: 20135875-20135875 on Build GRCh38
rs761048193 Chr.X: 20135887-20135887 on Build GRCh38
rs764897427 Chr.X: 20135867-20135867 on Build GRCh38
rs749301134 Chr.X: 20135864-20135864 on Build GRCh38
rs759798231 Chr.X: 20135756-20135756 on Build GRCh38
rs181821614 Chr.X: 20135847-20135847 on Build GRCh38
rs373076290 Chr.X: 20135889-20135889 on Build GRCh38
rs5955886 Chr.X: 20135846-20135846 on Build GRCh38
rs756358787 Chr.X: 20135862-20135862 on Build GRCh38
rs375192872 Chr.X: 20135861-20135861 on Build GRCh38
rs374795026 Chr.X: 20135686-20135686 on Build GRCh38
rs151115944 Chr.X: 20135786-20135786 on Build GRCh38
rs769987899 Chr.X: 20135695-20135695 on Build GRCh38
rs763748273 Chr.X: 20135806-20135806 on Build GRCh38
rs767982673 Chr.X: 20135779-20135779 on Build GRCh38
rs774893237 Chr.X: 20135886-20135887 on Build GRCh38
rs191689440 Chr.X: 20135680-20135680 on Build GRCh38
rs369630362 Chr.X: 20135863-20135863 on Build GRCh38
rs753293956 Chr.X: 20135837-20135837 on Build GRCh38
rs141026404 Chr.X: 20135795-20135795 on Build GRCh38
rs775772265 Chr.X: 20135881-20135881 on Build GRCh38
rs760524182 Chr.X: 20135702-20135702 on Build GRCh38
rs780059737 Chr.X: 20135856-20135856 on Build GRCh38
rs765842756 Chr.X: 20135683-20135683 on Build GRCh38
rs763354681 Chr.X: 20135888-20135888 on Build GRCh38
rs748437157 Chr.X: 20135690-20135690 on Build GRCh38
rs768571259 Chr.X: 20135733-20135733 on Build GRCh38
rs750044706 Chr.X: 20135870-20135870 on Build GRCh38

View Full Product Details