Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 105479137-105479382 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRIP2
Entrez Gene ID
1397
Gene Name
cysteine rich protein 2
Gene Aliases
CRIP, CRP2, ESP1
UniGene
Hs.534309
Gene Chromosome Location
Chr.14: 105472938-105480170 on Build GRCh38
Transcripts
NM_001270837.1 NM_001270841.1 NM_001312.3 NR_073081.1 NR_073082.1 NR_073083.1 NR_073084.1 NR_073085.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6391734 c.438G>A p.W146* Chr.14: 105479156-105479156 on Build GRCh38
4049572 c.459C>T p.C153C Chr.14: 105479177-105479177 on Build GRCh38
4800612 c.464G>C p.R155P Chr.14: 105479182-105479182 on Build GRCh38
697768 c.464G>T p.R155L Chr.14: 105479182-105479182 on Build GRCh38
5847715 c.485C>T p.P162L Chr.14: 105479203-105479203 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs781786654 Chr.14: 105479170-105479170 on Build GRCh38
rs782778280 Chr.14: 105479159-105479159 on Build GRCh38
rs782762445 Chr.14: 105479253-105479253 on Build GRCh38
rs782276346 Chr.14: 105479202-105479202 on Build GRCh38
rs1020015243 Chr.14: 105479362-105479362 on Build GRCh38
rs942540528 Chr.14: 105479272-105479272 on Build GRCh38
rs782237184 Chr.14: 105479230-105479230 on Build GRCh38
rs782160175 Chr.14: 105479240-105479240 on Build GRCh38
rs782132634 Chr.14: 105479252-105479252 on Build GRCh38
rs587651277 Chr.14: 105479304-105479304 on Build GRCh38
rs587665278 Chr.14: 105479348-105479348 on Build GRCh38
rs1035562263 Chr.14: 105479339-105479339 on Build GRCh38
rs922463911 Chr.14: 105479278-105479278 on Build GRCh38
rs587734882 Chr.14: 105479299-105479299 on Build GRCh38
rs959740616 Chr.14: 105479180-105479180 on Build GRCh38
rs887084802 Chr.14: 105479301-105479301 on Build GRCh38
rs202088808 Chr.14: 105479183-105479183 on Build GRCh38
rs975691685 Chr.14: 105479273-105479273 on Build GRCh38
rs782262017 Chr.14: 105479178-105479178 on Build GRCh38
rs978629484 Chr.14: 105479257-105479257 on Build GRCh38
rs925891188 Chr.14: 105479265-105479265 on Build GRCh38
rs781904647 Chr.14: 105479262-105479262 on Build GRCh38
rs782529567 Chr.14: 105479268-105479268 on Build GRCh38
rs782351211 Chr.14: 105479236-105479236 on Build GRCh38
rs11556117 Chr.14: 105479198-105479198 on Build GRCh38
rs782521914 Chr.14: 105479161-105479161 on Build GRCh38
rs587644713 Chr.14: 105479268-105479268 on Build GRCh38
rs782814440 Chr.14: 105479170-105479171 on Build GRCh38
rs992542614 Chr.14: 105479186-105479186 on Build GRCh38
rs941367049 Chr.14: 105479308-105479308 on Build GRCh38
rs145201995 Chr.14: 105479160-105479160 on Build GRCh38
rs1019877387 Chr.14: 105479219-105479219 on Build GRCh38
rs782485931 Chr.14: 105479177-105479177 on Build GRCh38
rs782047009 Chr.14: 105479248-105479248 on Build GRCh38
rs905478390 Chr.14: 105479335-105479335 on Build GRCh38
rs200032706 Chr.14: 105479157-105479157 on Build GRCh38
rs587632346 Chr.14: 105479327-105479327 on Build GRCh38
rs375650584 Chr.14: 105479165-105479165 on Build GRCh38
rs587602810 Chr.14: 105479246-105479246 on Build GRCh38
rs782326079 Chr.14: 105479207-105479207 on Build GRCh38
rs1034098714 Chr.14: 105479163-105479163 on Build GRCh38
rs782511415 Chr.14: 105479181-105479181 on Build GRCh38
rs781930160 Chr.14: 105479247-105479247 on Build GRCh38
rs1038607506 Chr.14: 105479334-105479334 on Build GRCh38
rs896670743 Chr.14: 105479341-105479341 on Build GRCh38
rs587679419 Chr.14: 105479293-105479293 on Build GRCh38
rs782009863 Chr.14: 105479239-105479239 on Build GRCh38
rs1002460344 Chr.14: 105479338-105479338 on Build GRCh38
rs748461792 Chr.14: 105479294-105479294 on Build GRCh38
rs782583716 Chr.14: 105479226-105479226 on Build GRCh38

View Full Product Details