Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 108510287-108510548 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXPH5
Entrez Gene ID
23086
Gene Name
exophilin 5
Gene Aliases
SLAC2-B, SLAC2B
UniGene
Hs.28540
Gene Chromosome Location
Chr.11: 108505431-108607514 on Build GRCh38
Transcripts
NM_001144763.1 NM_001144764.1 NM_001144765.1 NM_001308019.1 NM_015065.2 XM_011542696.2 XM_011542698.2 XM_011542700.2 XM_017017397.1 XM_017017398.1 XM_017017399.1 XM_017017400.1 XM_017017401.1 XM_017017402.1 XM_017017403.1 XM_017017404.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038521 c.5216G>T p.S1739I Chr.11: 108510291-108510291 on Build GRCh38
3985896 c.5208A>C p.T1736T Chr.11: 108510299-108510299 on Build GRCh38
6306989 c.5217C>A p.S1739R Chr.11: 108510290-108510290 on Build GRCh38
147321 c.4987G>A p.G1663R Chr.11: 108510520-108510520 on Build GRCh38
4574030 c.5154C>T p.D1718D Chr.11: 108510353-108510353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750387420 Chr.11: 108510316-108510316 on Build GRCh38
rs891233254 Chr.11: 108510515-108510515 on Build GRCh38
rs971652642 Chr.11: 108510343-108510343 on Build GRCh38
rs778459714 Chr.11: 108510438-108510438 on Build GRCh38
rs766836128 Chr.11: 108510411-108510411 on Build GRCh38
rs920434753 Chr.11: 108510375-108510375 on Build GRCh38
rs373115356 Chr.11: 108510318-108510318 on Build GRCh38
rs780378830 Chr.11: 108510340-108510340 on Build GRCh38
rs149024755 Chr.11: 108510418-108510418 on Build GRCh38
rs376510289 Chr.11: 108510414-108510414 on Build GRCh38
rs748908788 Chr.11: 108510352-108510352 on Build GRCh38
rs755280942 Chr.11: 108510415-108510415 on Build GRCh38
rs777435529 Chr.11: 108510351-108510351 on Build GRCh38
rs759548587 Chr.11: 108510307-108510307 on Build GRCh38
rs554556853 Chr.11: 108510356-108510356 on Build GRCh38
rs148791411 Chr.11: 108510309-108510309 on Build GRCh38
rs115033293 Chr.11: 108510448-108510448 on Build GRCh38
rs114391005 Chr.11: 108510449-108510449 on Build GRCh38
rs1047964837 Chr.11: 108510462-108510462 on Build GRCh38
rs755501292 Chr.11: 108510486-108510486 on Build GRCh38
rs971925259 Chr.11: 108510334-108510334 on Build GRCh38
rs114306563 Chr.11: 108510353-108510353 on Build GRCh38
rs36022102 Chr.11: 108510391-108510391 on Build GRCh38
rs775393660 Chr.11: 108510368-108510368 on Build GRCh38
rs758871486 Chr.11: 108510410-108510410 on Build GRCh38
rs374841213 Chr.11: 108510379-108510379 on Build GRCh38
rs748963069 Chr.11: 108510431-108510431 on Build GRCh38
rs756055412 Chr.11: 108510367-108510367 on Build GRCh38
rs2640779 Chr.11: 108510520-108510520 on Build GRCh38
rs961952211 Chr.11: 108510322-108510322 on Build GRCh38
rs930534489 Chr.11: 108510377-108510377 on Build GRCh38
rs116768370 Chr.11: 108510464-108510464 on Build GRCh38
rs760505565 Chr.11: 108510370-108510370 on Build GRCh38
rs775988355 Chr.11: 108510375-108510376 on Build GRCh38
rs764448147 Chr.11: 108510314-108510314 on Build GRCh38
rs201992262 Chr.11: 108510521-108510521 on Build GRCh38
rs778865772 Chr.11: 108510327-108510327 on Build GRCh38
rs35276439 Chr.11: 108510523-108510523 on Build GRCh38
rs762074616 Chr.11: 108510460-108510460 on Build GRCh38
rs756915127 Chr.11: 108510436-108510436 on Build GRCh38
rs186095969 Chr.11: 108510484-108510484 on Build GRCh38
rs1045420842 Chr.11: 108510442-108510442 on Build GRCh38
rs759742725 Chr.11: 108510373-108510374 on Build GRCh38
rs1037665956 Chr.11: 108510481-108510481 on Build GRCh38
rs767415161 Chr.11: 108510308-108510308 on Build GRCh38
rs758965358 Chr.11: 108510338-108510338 on Build GRCh38
rs144076188 Chr.11: 108510470-108510470 on Build GRCh38
rs755880369 Chr.11: 108510500-108510500 on Build GRCh38
rs1022772725 Chr.11: 108510329-108510329 on Build GRCh38
rs763815499 Chr.11: 108510378-108510378 on Build GRCh38
rs750551739 Chr.11: 108510402-108510402 on Build GRCh38
rs753049354 Chr.11: 108510499-108510499 on Build GRCh38
rs767906224 Chr.11: 108510374-108510375 on Build GRCh38
rs755367080 Chr.11: 108510348-108510348 on Build GRCh38
rs34026967 Chr.11: 108510451-108510451 on Build GRCh38
rs565716463 Chr.11: 108510320-108510320 on Build GRCh38
rs745407367 Chr.11: 108510445-108510445 on Build GRCh38
rs762975636 Chr.11: 108510479-108510479 on Build GRCh38
rs145299498 Chr.11: 108510362-108510362 on Build GRCh38
rs773458732 Chr.11: 108510471-108510471 on Build GRCh38
rs761161943 Chr.11: 108510474-108510478 on Build GRCh38
rs375603447 Chr.11: 108510494-108510494 on Build GRCh38
rs943595474 Chr.11: 108510426-108510426 on Build GRCh38
rs553059937 Chr.11: 108510476-108510476 on Build GRCh38
rs893389484 Chr.11: 108510466-108510466 on Build GRCh38
rs750179250 Chr.11: 108510440-108510440 on Build GRCh38
rs776349126 Chr.11: 108510457-108510457 on Build GRCh38
rs750938728 Chr.11: 108510331-108510331 on Build GRCh38
rs747413875 Chr.11: 108510346-108510346 on Build GRCh38
rs924348000 Chr.11: 108510369-108510369 on Build GRCh38

View Full Product Details