Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 131084773-131085016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGAP36
Entrez Gene ID
158763
Gene Name
Rho GTPase activating protein 36
Gene Aliases
-
UniGene
Hs.22905
Gene Chromosome Location
Chr.X: 131058242-131089885 on Build GRCh38
Transcripts
NM_001282607.1 NM_144967.3 XM_006724729.1 XM_011531280.1

SNP Detail

SNP ID Chromosome Location
rs760865099 Chr.X: 131084884-131084884 on Build GRCh38
rs992918417 Chr.X: 131084823-131084823 on Build GRCh38
rs752695138 Chr.X: 131084903-131084903 on Build GRCh38
rs749994409 Chr.X: 131084986-131084986 on Build GRCh38
rs368446020 Chr.X: 131084892-131084892 on Build GRCh38
rs202056670 Chr.X: 131084960-131084960 on Build GRCh38
rs766514700 Chr.X: 131084890-131084890 on Build GRCh38
rs369297346 Chr.X: 131084907-131084907 on Build GRCh38
rs776548814 Chr.X: 131084927-131084927 on Build GRCh38
rs762916133 Chr.X: 131084914-131084914 on Build GRCh38
rs755635983 Chr.X: 131084988-131084988 on Build GRCh38
rs780930341 Chr.X: 131084984-131084984 on Build GRCh38
rs201473039 Chr.X: 131084866-131084866 on Build GRCh38
rs751427111 Chr.X: 131084936-131084936 on Build GRCh38
rs772383550 Chr.X: 131084875-131084875 on Build GRCh38
rs140912896 Chr.X: 131084993-131084993 on Build GRCh38
rs773273631 Chr.X: 131084876-131084876 on Build GRCh38
rs758632079 Chr.X: 131084853-131084853 on Build GRCh38

View Full Product Details