Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 10710741-10710976 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADPRM
Entrez Gene ID
56985
Gene Name
ADP-ribose/CDP-alcohol diphosphatase, manganese dependent
Gene Aliases
C17orf48, MDS006, NBLA03831
UniGene
Hs.47668
Gene Chromosome Location
Chr.17: 10697610-10711558 on Build GRCh38
Transcripts
NM_020233.4 XM_005256738.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
135741 c.737C>T p.P246L Chr.17: 10710852-10710852 on Build GRCh38
5481340 c.741C>T p.D247D Chr.17: 10710856-10710856 on Build GRCh38
279300 c.822C>A p.F274L Chr.17: 10710937-10710937 on Build GRCh38
4619446 c.742G>A p.A248T Chr.17: 10710857-10710857 on Build GRCh38
5567956 c.853T>A p.S285T Chr.17: 10710968-10710968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775380575 Chr.17: 10710844-10710844 on Build GRCh38
rs371208948 Chr.17: 10710850-10710850 on Build GRCh38
rs750352845 Chr.17: 10710873-10710873 on Build GRCh38
rs761697635 Chr.17: 10710866-10710866 on Build GRCh38
rs750967584 Chr.17: 10710902-10710902 on Build GRCh38
rs891457562 Chr.17: 10710852-10710852 on Build GRCh38
rs765165765 Chr.17: 10710867-10710867 on Build GRCh38
rs752410674 Chr.17: 10710928-10710928 on Build GRCh38
rs1043354395 Chr.17: 10710865-10710865 on Build GRCh38
rs369400345 Chr.17: 10710800-10710800 on Build GRCh38
rs757584179 Chr.17: 10710949-10710949 on Build GRCh38
rs754527461 Chr.17: 10710910-10710910 on Build GRCh38
rs767142700 Chr.17: 10710922-10710922 on Build GRCh38
rs199523732 Chr.17: 10710857-10710857 on Build GRCh38
rs746400669 Chr.17: 10710816-10710816 on Build GRCh38
rs779855764 Chr.17: 10710943-10710944 on Build GRCh38
rs746262899 Chr.17: 10710953-10710953 on Build GRCh38
rs373696635 Chr.17: 10710853-10710853 on Build GRCh38
rs560251856 Chr.17: 10710887-10710887 on Build GRCh38
rs368408429 Chr.17: 10710856-10710856 on Build GRCh38
rs200237756 Chr.17: 10710788-10710788 on Build GRCh38
rs772165535 Chr.17: 10710839-10710839 on Build GRCh38
rs779294932 Chr.17: 10710951-10710951 on Build GRCh38
rs762751321 Chr.17: 10710885-10710885 on Build GRCh38
rs755015792 Chr.17: 10710787-10710787 on Build GRCh38

View Full Product Details