Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 17494844-17495069 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BFSP1
Entrez Gene ID
631
Gene Name
beaded filament structural protein 1
Gene Aliases
CP115, CP94, CTRCT33, LIFL-H
UniGene
Hs.129702
Gene Chromosome Location
Chr.20: 17493905-17569220 on Build GRCh38
Transcripts
NM_001161705.1 NM_001195.4 NM_001278606.1 NM_001278607.1 NM_001278608.1 XM_017028005.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3840497 c.685C>T p.Q229* Chr.20: 17495012-17495012 on Build GRCh38
4871218 c.729G>A p.K243K Chr.20: 17494968-17494968 on Build GRCh38
4665659 c.1150G>A p.G384R Chr.20: 17494922-17494922 on Build GRCh38
1636708 c.1209A>G p.V403V Chr.20: 17494863-17494863 on Build GRCh38
3544356 c.1149C>T p.N383N Chr.20: 17494923-17494923 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763821625 Chr.20: 17494883-17494883 on Build GRCh38
rs751346192 Chr.20: 17494895-17494895 on Build GRCh38
rs755188734 Chr.20: 17495009-17495009 on Build GRCh38
rs774430170 Chr.20: 17494964-17494964 on Build GRCh38
rs769207170 Chr.20: 17494946-17494946 on Build GRCh38
rs377289923 Chr.20: 17495003-17495003 on Build GRCh38
rs752093667 Chr.20: 17494903-17494903 on Build GRCh38
rs758874244 Chr.20: 17494923-17494923 on Build GRCh38
rs140116733 Chr.20: 17494878-17494878 on Build GRCh38
rs745495348 Chr.20: 17494940-17494940 on Build GRCh38
rs962809458 Chr.20: 17494879-17494879 on Build GRCh38
rs764369789 Chr.20: 17494882-17494890 on Build GRCh38
rs766776703 Chr.20: 17494904-17494904 on Build GRCh38
rs767400938 Chr.20: 17495002-17495002 on Build GRCh38
rs372521408 Chr.20: 17494938-17494938 on Build GRCh38
rs765236451 Chr.20: 17495013-17495013 on Build GRCh38
rs771279443 Chr.20: 17494937-17494937 on Build GRCh38
rs752763881 Chr.20: 17495014-17495014 on Build GRCh38
rs918268133 Chr.20: 17494909-17494909 on Build GRCh38
rs761742869 Chr.20: 17494972-17494972 on Build GRCh38
rs755063095 Chr.20: 17494910-17494910 on Build GRCh38
rs748635003 Chr.20: 17494922-17494922 on Build GRCh38
rs537903745 Chr.20: 17494886-17494886 on Build GRCh38
rs139124891 Chr.20: 17494947-17494947 on Build GRCh38
rs754085071 Chr.20: 17494907-17494907 on Build GRCh38
rs372928259 Chr.20: 17494949-17494949 on Build GRCh38
rs150798461 Chr.20: 17494924-17494924 on Build GRCh38
rs1022461977 Chr.20: 17494971-17494971 on Build GRCh38
rs747431829 Chr.20: 17494933-17494933 on Build GRCh38
rs775335712 Chr.20: 17494876-17494876 on Build GRCh38
rs754138243 Chr.20: 17495005-17495005 on Build GRCh38
rs779003224 Chr.20: 17494913-17494913 on Build GRCh38
rs143916769 Chr.20: 17494898-17494898 on Build GRCh38

View Full Product Details