Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 199961806-199962071 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf47
Entrez Gene ID
79568
Gene Name
chromosome 2 open reading frame 47
Gene Aliases
-
UniGene
Hs.154494
Gene Chromosome Location
Chr.2: 199955317-199964125 on Build GRCh38
Transcripts
NM_024520.2 XM_005246852.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4583078 c.725G>C p.G242A Chr.2: 199961856-199961856 on Build GRCh38
3838245 c.781C>A p.L261I Chr.2: 199961912-199961912 on Build GRCh38
2906221 c.701A>G p.N234S Chr.2: 199961832-199961832 on Build GRCh38
1482610 c.742G>T p.V248F Chr.2: 199961873-199961873 on Build GRCh38
5497371 c.752G>C p.G251A Chr.2: 199961883-199961883 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199942077 Chr.2: 199961963-199961963 on Build GRCh38
rs554788216 Chr.2: 199961937-199961937 on Build GRCh38
rs762689727 Chr.2: 199961977-199961977 on Build GRCh38
rs146994812 Chr.2: 199961928-199961928 on Build GRCh38
rs927455708 Chr.2: 199962045-199962045 on Build GRCh38
rs912857326 Chr.2: 199962028-199962028 on Build GRCh38
rs768716654 Chr.2: 199961933-199961933 on Build GRCh38
rs184949642 Chr.2: 199962033-199962033 on Build GRCh38
rs773849867 Chr.2: 199961949-199961949 on Build GRCh38
rs147805097 Chr.2: 199961841-199961841 on Build GRCh38
rs756203672 Chr.2: 199961922-199961922 on Build GRCh38
rs987033057 Chr.2: 199962025-199962025 on Build GRCh38
rs752410100 Chr.2: 199961832-199961832 on Build GRCh38
rs542762716 Chr.2: 199961923-199961923 on Build GRCh38
rs201762968 Chr.2: 199961932-199961932 on Build GRCh38
rs751307765 Chr.2: 199961869-199961869 on Build GRCh38
rs761030922 Chr.2: 199961955-199961955 on Build GRCh38
rs72928264 Chr.2: 199962014-199962014 on Build GRCh38
rs369693490 Chr.2: 199961971-199961971 on Build GRCh38
rs757238753 Chr.2: 199961870-199961870 on Build GRCh38
rs772744792 Chr.2: 199961948-199961948 on Build GRCh38
rs971688978 Chr.2: 199962043-199962043 on Build GRCh38
rs954263059 Chr.2: 199962024-199962024 on Build GRCh38
rs763635031 Chr.2: 199961980-199961980 on Build GRCh38
rs201092557 Chr.2: 199961902-199961902 on Build GRCh38
rs540716498 Chr.2: 199961986-199961986 on Build GRCh38
rs971355743 Chr.2: 199961916-199961916 on Build GRCh38
rs773223479 Chr.2: 199961955-199961955 on Build GRCh38
rs763981865 Chr.2: 199961850-199961850 on Build GRCh38
rs376452416 Chr.2: 199961906-199961906 on Build GRCh38
rs144184418 Chr.2: 199961830-199961830 on Build GRCh38
rs748500806 Chr.2: 199961942-199961942 on Build GRCh38
rs554643599 Chr.2: 199961847-199961848 on Build GRCh38
rs912238658 Chr.2: 199961873-199961873 on Build GRCh38

View Full Product Details