Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171337259-171337518 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO4
Entrez Gene ID
2329
Gene Name
flavin containing monooxygenase 4
Gene Aliases
FMO2
UniGene
Hs.386502
Gene Chromosome Location
Chr.1: 171314131-171342084 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
311246 c.1184T>C p.L395P Chr.1: 171337359-171337359 on Build GRCh38
4685160 c.1245T>C p.I415I Chr.1: 171337420-171337420 on Build GRCh38
424587 c.1219G>C p.E407Q Chr.1: 171337394-171337394 on Build GRCh38
6059600 c.1248A>G p.K416K Chr.1: 171337423-171337423 on Build GRCh38
5684323 c.1219G>T p.E407* Chr.1: 171337394-171337394 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767066955 Chr.1: 171337322-171337322 on Build GRCh38
rs752543666 Chr.1: 171337332-171337332 on Build GRCh38
rs917048718 Chr.1: 171337396-171337396 on Build GRCh38
rs765828660 Chr.1: 171337312-171337312 on Build GRCh38
rs746024147 Chr.1: 171337389-171337389 on Build GRCh38
rs780555012 Chr.1: 171337412-171337412 on Build GRCh38
rs1006212082 Chr.1: 171337333-171337333 on Build GRCh38
rs144924333 Chr.1: 171337397-171337397 on Build GRCh38
rs978385604 Chr.1: 171337313-171337313 on Build GRCh38
rs778641691 Chr.1: 171337374-171337375 on Build GRCh38
rs764915867 Chr.1: 171337467-171337467 on Build GRCh38
rs567230609 Chr.1: 171337487-171337487 on Build GRCh38
rs769013150 Chr.1: 171337426-171337426 on Build GRCh38
rs74962957 Chr.1: 171337331-171337331 on Build GRCh38
rs776188222 Chr.1: 171337448-171337448 on Build GRCh38
rs758305858 Chr.1: 171337341-171337341 on Build GRCh38
rs760272122 Chr.1: 171337310-171337310 on Build GRCh38
rs759173642 Chr.1: 171337465-171337465 on Build GRCh38
rs757795780 Chr.1: 171337474-171337475 on Build GRCh38
rs749966035 Chr.1: 171337459-171337459 on Build GRCh38
rs774872723 Chr.1: 171337433-171337433 on Build GRCh38
rs781148474 Chr.1: 171337385-171337385 on Build GRCh38
rs372467669 Chr.1: 171337347-171337347 on Build GRCh38
rs755513024 Chr.1: 171337449-171337449 on Build GRCh38
rs775230221 Chr.1: 171337470-171337470 on Build GRCh38
rs1034566829 Chr.1: 171337357-171337357 on Build GRCh38
rs35591720 Chr.1: 171337443-171337443 on Build GRCh38
rs757037297 Chr.1: 171337343-171337344 on Build GRCh38
rs749720900 Chr.1: 171337422-171337422 on Build GRCh38
rs748614732 Chr.1: 171337437-171337437 on Build GRCh38
rs905256530 Chr.1: 171337452-171337452 on Build GRCh38
rs191010605 Chr.1: 171337482-171337482 on Build GRCh38
rs776208286 Chr.1: 171337321-171337321 on Build GRCh38
rs751478744 Chr.1: 171337380-171337380 on Build GRCh38
rs45445497 Chr.1: 171337381-171337381 on Build GRCh38

View Full Product Details