Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5867395-5867648 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUT5
Entrez Gene ID
2527
Gene Name
fucosyltransferase 5
Gene Aliases
FUC-TV
UniGene
Hs.631843
Gene Chromosome Location
Chr.19: 5865826-5870540 on Build GRCh38
Transcripts
NM_002034.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4685895 c.302C>T p.A101V Chr.19: 5867424-5867424 on Build GRCh38
6667893 c.155T>C p.V52A Chr.19: 5867571-5867571 on Build GRCh38
1641263 c.187C>A p.R63S Chr.19: 5867539-5867539 on Build GRCh38
3540723 c.234C>T p.I78I Chr.19: 5867492-5867492 on Build GRCh38
3540724 c.207G>A p.A69A Chr.19: 5867519-5867519 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371381431 Chr.19: 5867547-5867547 on Build GRCh38
rs747185407 Chr.19: 5867539-5867539 on Build GRCh38
rs752344799 Chr.19: 5867507-5867507 on Build GRCh38
rs769062656 Chr.19: 5867543-5867543 on Build GRCh38
rs141146690 Chr.19: 5867538-5867538 on Build GRCh38
rs570071116 Chr.19: 5867608-5867608 on Build GRCh38
rs566802540 Chr.19: 5867442-5867442 on Build GRCh38
rs59903004 Chr.19: 5867555-5867555 on Build GRCh38
rs140732692 Chr.19: 5867424-5867424 on Build GRCh38
rs763786970 Chr.19: 5867569-5867569 on Build GRCh38
rs531111057 Chr.19: 5867609-5867609 on Build GRCh38
rs754840078 Chr.19: 5867502-5867502 on Build GRCh38
rs760504682 Chr.19: 5867618-5867618 on Build GRCh38
rs780946375 Chr.19: 5867444-5867444 on Build GRCh38
rs1031888668 Chr.19: 5867604-5867604 on Build GRCh38
rs758420902 Chr.19: 5867428-5867428 on Build GRCh38
rs148233280 Chr.19: 5867491-5867491 on Build GRCh38
rs748857115 Chr.19: 5867520-5867520 on Build GRCh38
rs147222504 Chr.19: 5867575-5867575 on Build GRCh38
rs776850558 Chr.19: 5867489-5867489 on Build GRCh38
rs760851573 Chr.19: 5867423-5867423 on Build GRCh38
rs868466988 Chr.19: 5867484-5867484 on Build GRCh38
rs142806538 Chr.19: 5867610-5867610 on Build GRCh38
rs767284783 Chr.19: 5867566-5867566 on Build GRCh38
rs536359180 Chr.19: 5867419-5867419 on Build GRCh38
rs61731563 Chr.19: 5867448-5867448 on Build GRCh38
rs867761443 Chr.19: 5867576-5867576 on Build GRCh38
rs763603623 Chr.19: 5867491-5867491 on Build GRCh38
rs200984833 Chr.19: 5867494-5867494 on Build GRCh38
rs766478311 Chr.19: 5867493-5867493 on Build GRCh38
rs138074178 Chr.19: 5867558-5867558 on Build GRCh38
rs374135977 Chr.19: 5867480-5867480 on Build GRCh38
rs141356302 Chr.19: 5867461-5867461 on Build GRCh38
rs778311756 Chr.19: 5867577-5867577 on Build GRCh38
rs200650200 Chr.19: 5867452-5867452 on Build GRCh38
rs779439585 Chr.19: 5867537-5867537 on Build GRCh38
rs774303037 Chr.19: 5867557-5867557 on Build GRCh38
rs373593937 Chr.19: 5867429-5867429 on Build GRCh38
rs773179073 Chr.19: 5867614-5867614 on Build GRCh38
rs866925823 Chr.19: 5867511-5867511 on Build GRCh38
rs764663021 Chr.19: 5867591-5867591 on Build GRCh38
rs370745789 Chr.19: 5867468-5867468 on Build GRCh38
rs552417219 Chr.19: 5867504-5867504 on Build GRCh38
rs760833851 Chr.19: 5867483-5867483 on Build GRCh38
rs189234701 Chr.19: 5867458-5867458 on Build GRCh38
rs145907945 Chr.19: 5867425-5867425 on Build GRCh38
rs775335134 Chr.19: 5867617-5867617 on Build GRCh38
rs767246713 Chr.19: 5867506-5867506 on Build GRCh38
rs865797115 Chr.19: 5867522-5867522 on Build GRCh38
rs763909943 Chr.19: 5867623-5867623 on Build GRCh38
rs1037131192 Chr.19: 5867476-5867476 on Build GRCh38
rs201359166 Chr.19: 5867584-5867584 on Build GRCh38
rs181646788 Chr.19: 5867519-5867519 on Build GRCh38
rs144634186 Chr.19: 5867481-5867481 on Build GRCh38
rs757741314 Chr.19: 5867526-5867526 on Build GRCh38
rs564509547 Chr.19: 5867572-5867572 on Build GRCh38
rs747433530 Chr.19: 5867606-5867606 on Build GRCh38
rs556886147 Chr.19: 5867463-5867463 on Build GRCh38
rs143701783 Chr.19: 5867567-5867567 on Build GRCh38
rs140463380 Chr.19: 5867598-5867598 on Build GRCh38
rs370008629 Chr.19: 5867550-5867550 on Build GRCh38
rs750888983 Chr.19: 5867583-5867583 on Build GRCh38
rs759514062 Chr.19: 5867616-5867616 on Build GRCh38
rs764208995 Chr.19: 5867486-5867486 on Build GRCh38
rs957230392 Chr.19: 5867607-5867607 on Build GRCh38
rs144284886 Chr.19: 5867613-5867613 on Build GRCh38

View Full Product Details