Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 49851888-49852131 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CRISP1
Entrez Gene ID
167
Gene Name
cysteine rich secretory protein 1
Gene Aliases
AEGL1, ARP, CRISP-1, HEL-S-57, HSCRISP1D, HSCRISP1G, HUMARP
UniGene
Hs.109620
Gene Chromosome Location
Chr.6: 49834257-49877096 on Build GRCh38
Transcripts
NM_001131.2 NM_001205220.1 NM_170609.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6862927 c.67-1G>A p.? Chr.6: 49852130-49852130 on Build GRCh38
4674718 c.91A>C p.N31H Chr.6: 49852105-49852105 on Build GRCh38
3628340 c.69G>A p.K23K Chr.6: 49852127-49852127 on Build GRCh38
6842277 c.86A>C p.Q29P Chr.6: 49852110-49852110 on Build GRCh38
6106893 c.176C>A p.A59D Chr.6: 49852020-49852020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373547128 Chr.6: 49851993-49851993 on Build GRCh38
rs760276316 Chr.6: 49851966-49851966 on Build GRCh38
rs966420383 Chr.6: 49852093-49852093 on Build GRCh38
rs997555064 Chr.6: 49852007-49852007 on Build GRCh38
rs750276251 Chr.6: 49851990-49851990 on Build GRCh38
rs187785377 Chr.6: 49852096-49852096 on Build GRCh38
rs753555800 Chr.6: 49851973-49851973 on Build GRCh38
rs150253145 Chr.6: 49852063-49852063 on Build GRCh38
rs142755504 Chr.6: 49852009-49852009 on Build GRCh38
rs780016479 Chr.6: 49851997-49851997 on Build GRCh38
rs958567346 Chr.6: 49852098-49852098 on Build GRCh38
rs145871288 Chr.6: 49852091-49852091 on Build GRCh38
rs537304746 Chr.6: 49852080-49852080 on Build GRCh38
rs755151900 Chr.6: 49852068-49852070 on Build GRCh38
rs748419084 Chr.6: 49852011-49852011 on Build GRCh38
rs773657772 Chr.6: 49851956-49851956 on Build GRCh38
rs373430755 Chr.6: 49852057-49852057 on Build GRCh38
rs758211722 Chr.6: 49852056-49852056 on Build GRCh38
rs771325857 Chr.6: 49851960-49851960 on Build GRCh38
rs763447011 Chr.6: 49852025-49852025 on Build GRCh38
rs1036846487 Chr.6: 49852017-49852017 on Build GRCh38
rs6915063 Chr.6: 49851959-49851959 on Build GRCh38
rs371336272 Chr.6: 49851971-49851971 on Build GRCh38
rs771395589 Chr.6: 49852028-49852028 on Build GRCh38
rs1006050957 Chr.6: 49852061-49852061 on Build GRCh38
rs759917401 Chr.6: 49852030-49852030 on Build GRCh38
rs147926729 Chr.6: 49852068-49852068 on Build GRCh38
rs775018778 Chr.6: 49852029-49852029 on Build GRCh38
rs764936134 Chr.6: 49852053-49852053 on Build GRCh38
rs867314509 Chr.6: 49852038-49852040 on Build GRCh38
rs756792397 Chr.6: 49851977-49851977 on Build GRCh38
rs774787737 Chr.6: 49852090-49852090 on Build GRCh38
rs770203243 Chr.6: 49851952-49851952 on Build GRCh38
rs151127841 Chr.6: 49852012-49852012 on Build GRCh38
rs763401642 Chr.6: 49851957-49851957 on Build GRCh38
rs370025572 Chr.6: 49852078-49852078 on Build GRCh38
rs778863581 Chr.6: 49852080-49852080 on Build GRCh38
rs549398921 Chr.6: 49851972-49851972 on Build GRCh38
rs749487003 Chr.6: 49851953-49851953 on Build GRCh38
rs761901702 Chr.6: 49851950-49851950 on Build GRCh38
rs925555445 Chr.6: 49851920-49851920 on Build GRCh38
rs753199391 Chr.6: 49852096-49852096 on Build GRCh38
rs776060725 Chr.6: 49852097-49852097 on Build GRCh38
rs773567666 Chr.6: 49852024-49852024 on Build GRCh38
rs763663914 Chr.6: 49852034-49852034 on Build GRCh38
rs768814191 Chr.6: 49852004-49852004 on Build GRCh38
rs937490216 Chr.6: 49851930-49851930 on Build GRCh38
rs139520539 Chr.6: 49852002-49852002 on Build GRCh38
rs201684630 Chr.6: 49852044-49852044 on Build GRCh38
rs141097725 Chr.6: 49852055-49852055 on Build GRCh38
rs199890851 Chr.6: 49852074-49852074 on Build GRCh38
rs751521013 Chr.6: 49852059-49852059 on Build GRCh38
rs778780372 Chr.6: 49851984-49851984 on Build GRCh38
rs563625923 Chr.6: 49851944-49851944 on Build GRCh38
rs753227426 Chr.6: 49852048-49852048 on Build GRCh38
rs141718389 Chr.6: 49852069-49852069 on Build GRCh38
rs971784181 Chr.6: 49852083-49852083 on Build GRCh38
rs376532347 Chr.6: 49852073-49852073 on Build GRCh38
rs761373495 Chr.6: 49852049-49852049 on Build GRCh38

View Full Product Details