Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 176392657-176392898 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARL10
Entrez Gene ID
285598
Gene Name
ADP ribosylation factor like GTPase 10
Gene Aliases
ARL10A
UniGene
Hs.424552
Gene Chromosome Location
Chr.5: 176365468-176415085 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001834.3 NM_007097.3 NR_045724.1 XM_017009026.1

Gene Symbol
CLTB
Entrez Gene ID
1212
Gene Name
clathrin light chain B
Gene Aliases
LCB
UniGene
Hs.484241
Gene Chromosome Location
Chr.5: 176392455-176416614 on Build GRCh38
Transcripts
XM_011534529.2 XM_011534530.2 NM_001834.3 NM_007097.3 NR_045724.1 XM_017009026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3702781 c.686G>A p.R229H Chr.5: 176392778-176392778 on Build GRCh38
4930604 c.656T>C p.L219P Chr.5: 176392808-176392808 on Build GRCh38
3853832 c.675G>A p.T225T Chr.5: 176392789-176392789 on Build GRCh38
4996541 c.575G>T p.W192L Chr.5: 176392889-176392889 on Build GRCh38
6630776 c.651G>A p.S217S Chr.5: 176392813-176392813 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs989929808 Chr.5: 176392759-176392769 on Build GRCh38
rs761220106 Chr.5: 176392789-176392789 on Build GRCh38
rs771241413 Chr.5: 176392778-176392778 on Build GRCh38
rs745895095 Chr.5: 176392780-176392780 on Build GRCh38
rs1019650291 Chr.5: 176392735-176392736 on Build GRCh38
rs544092722 Chr.5: 176392704-176392704 on Build GRCh38
rs764415111 Chr.5: 176392790-176392790 on Build GRCh38
rs928769518 Chr.5: 176392772-176392772 on Build GRCh38
rs79922429 Chr.5: 176392792-176392792 on Build GRCh38
rs201998729 Chr.5: 176392878-176392878 on Build GRCh38
rs1036351201 Chr.5: 176392680-176392680 on Build GRCh38
rs757983353 Chr.5: 176392725-176392725 on Build GRCh38
rs774449009 Chr.5: 176392779-176392779 on Build GRCh38
rs746961496 Chr.5: 176392733-176392733 on Build GRCh38
rs1031689656 Chr.5: 176392693-176392693 on Build GRCh38
rs145737672 Chr.5: 176392785-176392785 on Build GRCh38
rs761360140 Chr.5: 176392807-176392807 on Build GRCh38
rs374839373 Chr.5: 176392721-176392721 on Build GRCh38
rs563903618 Chr.5: 176392692-176392692 on Build GRCh38
rs372196026 Chr.5: 176392751-176392751 on Build GRCh38
rs966125067 Chr.5: 176392682-176392682 on Build GRCh38
rs902082988 Chr.5: 176392755-176392755 on Build GRCh38
rs111658327 Chr.5: 176392734-176392734 on Build GRCh38
rs770867733 Chr.5: 176392749-176392750 on Build GRCh38
rs764888931 Chr.5: 176392813-176392813 on Build GRCh38
rs768545653 Chr.5: 176392742-176392742 on Build GRCh38
rs780097314 Chr.5: 176392729-176392729 on Build GRCh38
rs1007810099 Chr.5: 176392726-176392726 on Build GRCh38
rs897935351 Chr.5: 176392685-176392685 on Build GRCh38
rs757930059 Chr.5: 176392818-176392818 on Build GRCh38
rs766030689 Chr.5: 176392866-176392866 on Build GRCh38
rs750071352 Chr.5: 176392814-176392814 on Build GRCh38
rs749466092 Chr.5: 176392766-176392766 on Build GRCh38
rs576020935 Chr.5: 176392695-176392695 on Build GRCh38
rs914847658 Chr.5: 176392750-176392750 on Build GRCh38

View Full Product Details