Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 222630034-222630281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FARSB
Entrez Gene ID
10056
Gene Name
phenylalanyl-tRNA synthetase beta subunit
Gene Aliases
FARSLB, FRSB, HSPC173, PheHB, PheRS
UniGene
Hs.471452
Gene Chromosome Location
Chr.2: 222571443-222656355 on Build GRCh38
Transcripts
NM_005687.4 NR_130154.1 XM_006712169.2 XM_011510466.2 XM_017003110.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6341129 c.816C>A p.T272T Chr.2: 222630145-222630145 on Build GRCh38
1017032 c.793A>G p.I265V Chr.2: 222630168-222630168 on Build GRCh38
6897669 c.796G>A p.V266I Chr.2: 222630165-222630165 on Build GRCh38
4929884 c.794T>C p.I265T Chr.2: 222630167-222630167 on Build GRCh38
6660624 c.799C>A p.L267I Chr.2: 222630162-222630162 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs979859908 Chr.2: 222630224-222630224 on Build GRCh38
rs755964857 Chr.2: 222630198-222630198 on Build GRCh38
rs771136862 Chr.2: 222630103-222630103 on Build GRCh38
rs377151075 Chr.2: 222630114-222630114 on Build GRCh38
rs778914283 Chr.2: 222630068-222630068 on Build GRCh38
rs758541288 Chr.2: 222630203-222630203 on Build GRCh38
rs996221829 Chr.2: 222630087-222630087 on Build GRCh38
rs749000123 Chr.2: 222630179-222630183 on Build GRCh38
rs762598366 Chr.2: 222630129-222630129 on Build GRCh38
rs767158624 Chr.2: 222630194-222630194 on Build GRCh38
rs938552853 Chr.2: 222630237-222630237 on Build GRCh38
rs759915281 Chr.2: 222630113-222630113 on Build GRCh38
rs370297363 Chr.2: 222630084-222630084 on Build GRCh38
rs748258257 Chr.2: 222630077-222630077 on Build GRCh38
rs752760514 Chr.2: 222630199-222630199 on Build GRCh38
rs376959679 Chr.2: 222630079-222630079 on Build GRCh38
rs1020998547 Chr.2: 222630133-222630133 on Build GRCh38
rs370583223 Chr.2: 222630197-222630197 on Build GRCh38
rs775992830 Chr.2: 222630230-222630230 on Build GRCh38
rs751321760 Chr.2: 222630137-222630137 on Build GRCh38
rs573531425 Chr.2: 222630246-222630246 on Build GRCh38
rs761529228 Chr.2: 222630162-222630162 on Build GRCh38
rs777836219 Chr.2: 222630221-222630221 on Build GRCh38
rs1019417502 Chr.2: 222630233-222630233 on Build GRCh38
rs961021002 Chr.2: 222630146-222630146 on Build GRCh38
rs984089178 Chr.2: 222630110-222630110 on Build GRCh38
rs778029153 Chr.2: 222630080-222630080 on Build GRCh38
rs775104835 Chr.2: 222630115-222630115 on Build GRCh38
rs954916476 Chr.2: 222630100-222630100 on Build GRCh38
rs755628041 Chr.2: 222630096-222630112 on Build GRCh38
rs777071414 Chr.2: 222630112-222630112 on Build GRCh38
rs182982049 Chr.2: 222630066-222630066 on Build GRCh38
rs372654250 Chr.2: 222630095-222630095 on Build GRCh38
rs770755314 Chr.2: 222630207-222630212 on Build GRCh38
rs763822806 Chr.2: 222630131-222630131 on Build GRCh38
rs757054935 Chr.2: 222630062-222630062 on Build GRCh38
rs187579379 Chr.2: 222630090-222630090 on Build GRCh38
rs756089922 Chr.2: 222630067-222630067 on Build GRCh38
rs777350879 Chr.2: 222630142-222630142 on Build GRCh38
rs1009967772 Chr.2: 222630124-222630124 on Build GRCh38

View Full Product Details