Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 42189574-42189827 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHDC
Entrez Gene ID
84514
Gene Name
GH3 domain containing
Gene Aliases
D11LGP1, LGP1
UniGene
Hs.38039
Gene Chromosome Location
Chr.17: 42189087-42194532 on Build GRCh38
Transcripts
NM_001142623.1 NM_032484.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6146439 c.1526C>A p.P509H Chr.17: 42189770-42189770 on Build GRCh38
4467914 c.1514C>T p.S505F Chr.17: 42189782-42189782 on Build GRCh38
5591107 c.1543C>T p.L515F Chr.17: 42189753-42189753 on Build GRCh38
6523539 c.1497C>T p.L499L Chr.17: 42189799-42189799 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs972319592 Chr.17: 42189773-42189773 on Build GRCh38
rs555615336 Chr.17: 42189733-42189733 on Build GRCh38
rs770309424 Chr.17: 42189714-42189714 on Build GRCh38
rs376472900 Chr.17: 42189798-42189798 on Build GRCh38
rs774695863 Chr.17: 42189644-42189644 on Build GRCh38
rs370000451 Chr.17: 42189810-42189810 on Build GRCh38
rs369559035 Chr.17: 42189711-42189711 on Build GRCh38
rs764389308 Chr.17: 42189654-42189654 on Build GRCh38
rs773811474 Chr.17: 42189652-42189652 on Build GRCh38
rs566800716 Chr.17: 42189658-42189658 on Build GRCh38
rs150943423 Chr.17: 42189715-42189715 on Build GRCh38
rs754155016 Chr.17: 42189702-42189702 on Build GRCh38
rs780198489 Chr.17: 42189789-42189789 on Build GRCh38
rs201510371 Chr.17: 42189771-42189771 on Build GRCh38
rs766276257 Chr.17: 42189672-42189672 on Build GRCh38
rs766336599 Chr.17: 42189750-42189750 on Build GRCh38
rs749656683 Chr.17: 42189721-42189721 on Build GRCh38
rs545336937 Chr.17: 42189767-42189767 on Build GRCh38
rs923367594 Chr.17: 42189749-42189749 on Build GRCh38
rs779394083 Chr.17: 42189809-42189809 on Build GRCh38
rs896214466 Chr.17: 42189635-42189635 on Build GRCh38
rs140765209 Chr.17: 42189759-42189759 on Build GRCh38
rs749988474 Chr.17: 42189671-42189671 on Build GRCh38
rs567813826 Chr.17: 42189740-42189740 on Build GRCh38
rs761521707 Chr.17: 42189734-42189734 on Build GRCh38
rs768847579 Chr.17: 42189730-42189730 on Build GRCh38
rs975911234 Chr.17: 42189731-42189731 on Build GRCh38
rs756212657 Chr.17: 42189788-42189788 on Build GRCh38
rs749713059 Chr.17: 42189797-42189797 on Build GRCh38
rs778772228 Chr.17: 42189674-42189674 on Build GRCh38
rs760319655 Chr.17: 42189743-42189743 on Build GRCh38
rs538074219 Chr.17: 42189758-42189758 on Build GRCh38
rs3744485 Chr.17: 42189656-42189656 on Build GRCh38
rs577928619 Chr.17: 42189766-42189766 on Build GRCh38
rs553704866 Chr.17: 42189769-42189769 on Build GRCh38
rs771894391 Chr.17: 42189739-42189739 on Build GRCh38
rs956776571 Chr.17: 42189716-42189716 on Build GRCh38
rs969915433 Chr.17: 42189792-42189792 on Build GRCh38
rs372222401 Chr.17: 42189598-42189598 on Build GRCh38
rs992617259 Chr.17: 42189775-42189775 on Build GRCh38
rs1022838679 Chr.17: 42189778-42189778 on Build GRCh38
rs934164642 Chr.17: 42189689-42189689 on Build GRCh38
rs897254809 Chr.17: 42189738-42189738 on Build GRCh38
rs761946797 Chr.17: 42189778-42189778 on Build GRCh38
rs750486249 Chr.17: 42189781-42189781 on Build GRCh38
rs765258680 Chr.17: 42189793-42189810 on Build GRCh38
rs559027453 Chr.17: 42189623-42189623 on Build GRCh38
rs757491741 Chr.17: 42189685-42189685 on Build GRCh38
rs751721448 Chr.17: 42189676-42189676 on Build GRCh38

View Full Product Details