Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 55874885-55875126 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFEMP1
Entrez Gene ID
2202
Gene Name
EGF containing fibulin like extracellular matrix protein 1
Gene Aliases
DHRD, DRAD, FBLN3, FBNL, FIBL-3, MLVT, MTLV, S1-5
UniGene
Hs.732348
Gene Chromosome Location
Chr.2: 55865962-55924163 on Build GRCh38
Transcripts
NM_001039348.2 NM_001039349.2 XM_005264205.4 XM_017003586.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
266444 c.928G>A p.V310I Chr.2: 55875018-55875018 on Build GRCh38
106484 c.989G>A p.R330K Chr.2: 55874957-55874957 on Build GRCh38
721467 c.983G>C p.R328T Chr.2: 55874963-55874963 on Build GRCh38
6040211 c.976G>T p.V326L Chr.2: 55874970-55874970 on Build GRCh38
5484553 c.931A>G p.N311D Chr.2: 55875015-55875015 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs770999988 Chr.2: 55874965-55874965 on Build GRCh38
rs764405236 Chr.2: 55875079-55875079 on Build GRCh38
rs777636747 Chr.2: 55875096-55875096 on Build GRCh38
rs777424675 Chr.2: 55874959-55874959 on Build GRCh38
rs150399072 Chr.2: 55874957-55874957 on Build GRCh38
rs373857382 Chr.2: 55875062-55875062 on Build GRCh38
rs912823881 Chr.2: 55874998-55874998 on Build GRCh38
rs774525572 Chr.2: 55875024-55875024 on Build GRCh38
rs925479971 Chr.2: 55874986-55874986 on Build GRCh38
rs367687928 Chr.2: 55875081-55875081 on Build GRCh38
rs187280049 Chr.2: 55875090-55875090 on Build GRCh38
rs776713948 Chr.2: 55874970-55874970 on Build GRCh38
rs796268760 Chr.2: 55874936-55874936 on Build GRCh38
rs752796496 Chr.2: 55874926-55874927 on Build GRCh38
rs753354829 Chr.2: 55874933-55874933 on Build GRCh38
rs769842736 Chr.2: 55874983-55874983 on Build GRCh38
rs146846635 Chr.2: 55875022-55875022 on Build GRCh38
rs564326837 Chr.2: 55874934-55874934 on Build GRCh38
rs76956725 Chr.2: 55874936-55874936 on Build GRCh38
rs988854831 Chr.2: 55875014-55875014 on Build GRCh38
rs748009744 Chr.2: 55874938-55874938 on Build GRCh38
rs745988780 Chr.2: 55874980-55874980 on Build GRCh38
rs747258798 Chr.2: 55874963-55874963 on Build GRCh38
rs148177279 Chr.2: 55874992-55874992 on Build GRCh38
rs544568752 Chr.2: 55874920-55874920 on Build GRCh38
rs886056191 Chr.2: 55874937-55874937 on Build GRCh38
rs766004890 Chr.2: 55874927-55874927 on Build GRCh38
rs540612217 Chr.2: 55874943-55874943 on Build GRCh38
rs143083501 Chr.2: 55875034-55875034 on Build GRCh38
rs759163094 Chr.2: 55875070-55875070 on Build GRCh38
rs755659448 Chr.2: 55874922-55874922 on Build GRCh38
rs965470528 Chr.2: 55874971-55874971 on Build GRCh38
rs374256179 Chr.2: 55875052-55875052 on Build GRCh38
rs979557875 Chr.2: 55874912-55874912 on Build GRCh38
rs954231474 Chr.2: 55874989-55874989 on Build GRCh38
rs556410164 Chr.2: 55875019-55875019 on Build GRCh38
rs760860820 Chr.2: 55874927-55874928 on Build GRCh38
rs764824404 Chr.2: 55875080-55875080 on Build GRCh38
rs750158924 Chr.2: 55874921-55874921 on Build GRCh38
rs75175808 Chr.2: 55874937-55874937 on Build GRCh38
rs763367920 Chr.2: 55874991-55874991 on Build GRCh38
rs56280335 Chr.2: 55874927-55874927 on Build GRCh38

View Full Product Details