Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172553037-172553310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SUCO
Entrez Gene ID
51430
Gene Name
SUN domain containing ossification factor
Gene Aliases
C1orf9, CH1, OPT, SLP1
UniGene
Hs.204559
Gene Chromosome Location
Chr.1: 172532349-172611833 on Build GRCh38
Transcripts
NM_001282750.1 NM_001282751.1 NM_014283.4 NM_016227.3 XM_006711374.2 XM_006711375.2 XM_006711376.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
899643 c.205G>A p.E69K Chr.1: 172553287-172553287 on Build GRCh38
3478391 c.221C>T p.S74L Chr.1: 172553303-172553303 on Build GRCh38
4763578 c.208T>C p.S70P Chr.1: 172553290-172553290 on Build GRCh38
4025115 c.180T>C p.D60D Chr.1: 172553262-172553262 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs914158474 Chr.1: 172553278-172553278 on Build GRCh38
rs1018742494 Chr.1: 172553068-172553068 on Build GRCh38
rs529654703 Chr.1: 172553114-172553114 on Build GRCh38
rs746886933 Chr.1: 172553281-172553281 on Build GRCh38
rs1027698007 Chr.1: 172553171-172553171 on Build GRCh38
rs764506470 Chr.1: 172553214-172553214 on Build GRCh38
rs757852561 Chr.1: 172553228-172553228 on Build GRCh38
rs766194690 Chr.1: 172553240-172553242 on Build GRCh38
rs777431044 Chr.1: 172553277-172553277 on Build GRCh38
rs571027148 Chr.1: 172553063-172553063 on Build GRCh38
rs979866767 Chr.1: 172553147-172553147 on Build GRCh38
rs17370159 Chr.1: 172553109-172553109 on Build GRCh38
rs960936225 Chr.1: 172553253-172553253 on Build GRCh38
rs548294334 Chr.1: 172553179-172553179 on Build GRCh38
rs540919651 Chr.1: 172553088-172553088 on Build GRCh38
rs530487523 Chr.1: 172553259-172553259 on Build GRCh38
rs1033873039 Chr.1: 172553245-172553245 on Build GRCh38
rs935417890 Chr.1: 172553178-172553179 on Build GRCh38
rs747880178 Chr.1: 172553262-172553262 on Build GRCh38
rs897795424 Chr.1: 172553124-172553124 on Build GRCh38
rs898764990 Chr.1: 172553276-172553276 on Build GRCh38
rs372922378 Chr.1: 172553268-172553268 on Build GRCh38
rs34585959 Chr.1: 172553179-172553179 on Build GRCh38
rs879127534 Chr.1: 172553157-172553158 on Build GRCh38
rs952144206 Chr.1: 172553193-172553193 on Build GRCh38
rs772142826 Chr.1: 172553099-172553099 on Build GRCh38
rs550102185 Chr.1: 172553147-172553150 on Build GRCh38
rs1029359551 Chr.1: 172553115-172553115 on Build GRCh38
rs754649444 Chr.1: 172553255-172553255 on Build GRCh38
rs560864714 Chr.1: 172553194-172553194 on Build GRCh38
rs142323342 Chr.1: 172553286-172553286 on Build GRCh38
rs765943396 Chr.1: 172553239-172553239 on Build GRCh38
rs202071817 Chr.1: 172553248-172553248 on Build GRCh38
rs111583627 Chr.1: 172553074-172553076 on Build GRCh38
rs993921692 Chr.1: 172553125-172553125 on Build GRCh38
rs753434649 Chr.1: 172553240-172553240 on Build GRCh38
rs752038333 Chr.1: 172553216-172553216 on Build GRCh38

View Full Product Details