Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 48918923-48919156 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARIH2
Entrez Gene ID
10425
Gene Name
ariadne RBR E3 ubiquitin protein ligase 2
Gene Aliases
ARI2, TRIAD1
UniGene
Hs.633601
Gene Chromosome Location
Chr.3: 48918820-48985541 on Build GRCh38
Transcripts
NM_001317333.1 NM_001317334.1 NM_006321.3 XM_011533265.1 XM_011533266.1 XM_011533267.1 XM_011533268.1 XM_011533270.1 XM_011533271.1 XM_011533273.1 XM_011533275.2 XM_017005531.1 XM_017005532.1 XM_017005534.1 XM_017005536.1 XM_017005537.1 XM_017005538.1 XM_017005539.1 XM_017005540.1 XM_017005541.1 NM_001123040.1

Gene Symbol
ARIH2OS
Entrez Gene ID
646450
Gene Name
ariadne homolog 2 opposite strand
Gene Aliases
C3orf71
UniGene
Hs.720727
Gene Chromosome Location
Chr.3: 48917788-48919385 on Build GRCh38
Transcripts
NM_001317333.1 NM_001317334.1 NM_006321.3 XM_011533265.1 XM_011533266.1 XM_011533267.1 XM_011533268.1 XM_011533270.1 XM_011533271.1 XM_011533273.1 XM_011533275.2 XM_017005531.1 XM_017005532.1 XM_017005534.1 XM_017005536.1 XM_017005537.1 XM_017005538.1 XM_017005539.1 XM_017005540.1 XM_017005541.1 NM_001123040.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4966477 c.62G>A p.G21E Chr.3: 48919088-48919088 on Build GRCh38
4667747 c.217_218insG p.D73fs*62 Chr.3: 48918932-48918933 on Build GRCh38
6502890 c.183G>T p.P61P Chr.3: 48918967-48918967 on Build GRCh38
6391654 c.142G>A p.A48T Chr.3: 48919008-48919008 on Build GRCh38
1742798 c.89C>A p.A30E Chr.3: 48919061-48919061 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1044821101 Chr.3: 48919017-48919017 on Build GRCh38
rs752659690 Chr.3: 48919023-48919023 on Build GRCh38
rs922299992 Chr.3: 48919051-48919051 on Build GRCh38
rs770729764 Chr.3: 48918993-48918993 on Build GRCh38
rs947949589 Chr.3: 48919074-48919074 on Build GRCh38
rs767308189 Chr.3: 48919010-48919010 on Build GRCh38
rs976961841 Chr.3: 48919019-48919019 on Build GRCh38
rs781595161 Chr.3: 48919117-48919117 on Build GRCh38
rs747564744 Chr.3: 48918967-48918967 on Build GRCh38
rs536644096 Chr.3: 48918987-48918987 on Build GRCh38
rs957495001 Chr.3: 48918975-48918975 on Build GRCh38
rs777776753 Chr.3: 48919131-48919131 on Build GRCh38
rs373018609 Chr.3: 48919101-48919101 on Build GRCh38
rs780190324 Chr.3: 48919092-48919092 on Build GRCh38
rs763150173 Chr.3: 48918989-48918989 on Build GRCh38
rs570126354 Chr.3: 48919102-48919102 on Build GRCh38
rs763691583 Chr.3: 48919079-48919079 on Build GRCh38
rs781359004 Chr.3: 48918968-48918968 on Build GRCh38
rs751826604 Chr.3: 48919106-48919106 on Build GRCh38
rs753600517 Chr.3: 48919090-48919090 on Build GRCh38
rs748125823 Chr.3: 48918970-48918970 on Build GRCh38
rs760144328 Chr.3: 48919075-48919075 on Build GRCh38
rs909582709 Chr.3: 48919053-48919053 on Build GRCh38
rs994527584 Chr.3: 48919129-48919129 on Build GRCh38
rs769762472 Chr.3: 48919007-48919007 on Build GRCh38
rs542334811 Chr.3: 48919097-48919123 on Build GRCh38
rs769838601 Chr.3: 48918973-48918973 on Build GRCh38
rs891896270 Chr.3: 48918949-48918949 on Build GRCh38
rs767060066 Chr.3: 48918943-48918943 on Build GRCh38
rs752109280 Chr.3: 48918947-48918947 on Build GRCh38
rs1024737553 Chr.3: 48918942-48918942 on Build GRCh38
rs749556955 Chr.3: 48919109-48919110 on Build GRCh38
rs537647405 Chr.3: 48919133-48919133 on Build GRCh38
rs372570123 Chr.3: 48918953-48918953 on Build GRCh38
rs990333468 Chr.3: 48918976-48918976 on Build GRCh38
rs879431590 Chr.3: 48918984-48918984 on Build GRCh38
rs548743370 Chr.3: 48919065-48919065 on Build GRCh38
rs759550722 Chr.3: 48919005-48919005 on Build GRCh38
rs946167681 Chr.3: 48919130-48919130 on Build GRCh38
rs769922215 Chr.3: 48919124-48919124 on Build GRCh38
rs550168944 Chr.3: 48918954-48918954 on Build GRCh38
rs754796075 Chr.3: 48919009-48919010 on Build GRCh38
rs1025794372 Chr.3: 48919024-48919024 on Build GRCh38
rs774069479 Chr.3: 48918998-48918998 on Build GRCh38
rs780552957 Chr.3: 48918961-48918961 on Build GRCh38
rs983775946 Chr.3: 48919048-48919048 on Build GRCh38
rs933736113 Chr.3: 48919064-48919064 on Build GRCh38
rs748917418 Chr.3: 48918957-48918957 on Build GRCh38
rs946298428 Chr.3: 48918997-48918997 on Build GRCh38
rs755528885 Chr.3: 48919111-48919111 on Build GRCh38
rs889546216 Chr.3: 48919098-48919098 on Build GRCh38
rs777957265 Chr.3: 48919046-48919047 on Build GRCh38
rs748687160 Chr.3: 48919122-48919122 on Build GRCh38

View Full Product Details