Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 121371251-121371484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM3C
Entrez Gene ID
10447
Gene Name
family with sequence similarity 3 member C
Gene Aliases
GS3786, ILEI
UniGene
Hs.434053
Gene Chromosome Location
Chr.7: 121348851-121396369 on Build GRCh38
Transcripts
NM_001040020.1 NM_014888.2 XM_011515736.2 XM_011515737.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2766470 c.264A>G p.E88E Chr.7: 121371308-121371308 on Build GRCh38
375878 c.177G>A p.G59G Chr.7: 121371395-121371395 on Build GRCh38
452216 c.237C>T p.N79N Chr.7: 121371335-121371335 on Build GRCh38
6108382 c.227G>C p.G76A Chr.7: 121371345-121371345 on Build GRCh38
6366219 c.241G>A p.V81M Chr.7: 121371331-121371331 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140177929 Chr.7: 121371397-121371397 on Build GRCh38
rs374629285 Chr.7: 121371380-121371380 on Build GRCh38
rs756222832 Chr.7: 121371448-121371449 on Build GRCh38
rs1010644782 Chr.7: 121371429-121371429 on Build GRCh38
rs748046331 Chr.7: 121371301-121371301 on Build GRCh38
rs758234623 Chr.7: 121371446-121371446 on Build GRCh38
rs778700686 Chr.7: 121371299-121371299 on Build GRCh38
rs771111364 Chr.7: 121371320-121371320 on Build GRCh38
rs143912709 Chr.7: 121371404-121371404 on Build GRCh38
rs778591929 Chr.7: 121371424-121371424 on Build GRCh38
rs776785879 Chr.7: 121371330-121371330 on Build GRCh38
rs759736620 Chr.7: 121371332-121371332 on Build GRCh38
rs950767093 Chr.7: 121371385-121371385 on Build GRCh38
rs747125032 Chr.7: 121371317-121371317 on Build GRCh38
rs752499174 Chr.7: 121371443-121371443 on Build GRCh38
rs773455264 Chr.7: 121371310-121371310 on Build GRCh38
rs201421824 Chr.7: 121371368-121371368 on Build GRCh38
rs763485264 Chr.7: 121371355-121371355 on Build GRCh38
rs200888829 Chr.7: 121371407-121371407 on Build GRCh38
rs775876766 Chr.7: 121371353-121371353 on Build GRCh38
rs765535690 Chr.7: 121371334-121371334 on Build GRCh38
rs376471348 Chr.7: 121371287-121371287 on Build GRCh38
rs753433677 Chr.7: 121371405-121371405 on Build GRCh38
rs755683116 Chr.7: 121371399-121371399 on Build GRCh38
rs772084430 Chr.7: 121371304-121371304 on Build GRCh38
rs957781500 Chr.7: 121371418-121371418 on Build GRCh38

View Full Product Details