Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 46058186-46058417 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPB2
Entrez Gene ID
1361
Gene Name
carboxypeptidase B2
Gene Aliases
CPU, PCPB, TAFI
UniGene
Hs.512937
Gene Chromosome Location
Chr.13: 46053186-46105076 on Build GRCh38
Transcripts
NM_001278541.1 NM_001872.4 XM_017020393.1 NR_046226.1 NR_046227.1

Gene Symbol
CPB2-AS1
Entrez Gene ID
100509894
Gene Name
CPB2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.626139
Gene Chromosome Location
Chr.13: 46052848-46101347 on Build GRCh38
Transcripts
NM_001278541.1 NM_001872.4 XM_017020393.1 NR_046226.1 NR_046227.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747462 c.947T>C p.I316T Chr.13: 46058231-46058231 on Build GRCh38
6634478 c.970C>T p.R324* Chr.13: 46058208-46058208 on Build GRCh38
4047855 c.854C>T p.S285L Chr.13: 46058324-46058324 on Build GRCh38
3731856 c.797-7G>A p.? Chr.13: 46058388-46058388 on Build GRCh38
4575953 c.826G>T p.E276* Chr.13: 46058352-46058352 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542442684 Chr.13: 46058365-46058365 on Build GRCh38
rs185905588 Chr.13: 46058239-46058239 on Build GRCh38
rs564181491 Chr.13: 46058280-46058280 on Build GRCh38
rs779579021 Chr.13: 46058305-46058305 on Build GRCh38
rs558696589 Chr.13: 46058272-46058272 on Build GRCh38
rs577140119 Chr.13: 46058275-46058275 on Build GRCh38
rs754395886 Chr.13: 46058296-46058296 on Build GRCh38
rs139468136 Chr.13: 46058334-46058334 on Build GRCh38
rs576834540 Chr.13: 46058219-46058219 on Build GRCh38
rs777244269 Chr.13: 46058351-46058351 on Build GRCh38
rs767565541 Chr.13: 46058274-46058274 on Build GRCh38
rs771568397 Chr.13: 46058225-46058225 on Build GRCh38
rs776528111 Chr.13: 46058259-46058259 on Build GRCh38
rs745791810 Chr.13: 46058377-46058377 on Build GRCh38
rs759509065 Chr.13: 46058262-46058262 on Build GRCh38
rs932147911 Chr.13: 46058354-46058354 on Build GRCh38
rs982716199 Chr.13: 46058231-46058231 on Build GRCh38
rs766611053 Chr.13: 46058288-46058288 on Build GRCh38
rs147810684 Chr.13: 46058353-46058353 on Build GRCh38
rs765011207 Chr.13: 46058263-46058263 on Build GRCh38
rs559840133 Chr.13: 46058284-46058284 on Build GRCh38
rs574588968 Chr.13: 46058363-46058363 on Build GRCh38
rs149654876 Chr.13: 46058347-46058347 on Build GRCh38
rs947994725 Chr.13: 46058308-46058308 on Build GRCh38
rs772924574 Chr.13: 46058232-46058232 on Build GRCh38
rs770499413 Chr.13: 46058245-46058245 on Build GRCh38
rs369983289 Chr.13: 46058388-46058388 on Build GRCh38
rs756784514 Chr.13: 46058369-46058370 on Build GRCh38
rs1046358643 Chr.13: 46058313-46058313 on Build GRCh38
rs769659242 Chr.13: 46058380-46058380 on Build GRCh38
rs907897258 Chr.13: 46058252-46058252 on Build GRCh38
rs755604530 Chr.13: 46058297-46058297 on Build GRCh38

View Full Product Details