Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49395742-49395969 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC155
Entrez Gene ID
147872
Gene Name
coiled-coil domain containing 155
Gene Aliases
KASH5
UniGene
Hs.662085
Gene Chromosome Location
Chr.19: 49388218-49418002 on Build GRCh38
Transcripts
NM_144688.4 XM_005258549.3 XM_011526488.2 XM_011526489.2 XM_011526490.2 XM_011526494.2 XM_011526495.2 XM_011526496.2 XM_011526497.2 XM_017026333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2752479 c.355G>C p.E119Q Chr.19: 49395788-49395788 on Build GRCh38
307677 c.361G>A p.A121T Chr.19: 49395794-49395794 on Build GRCh38
5002021 c.386G>A p.R129Q Chr.19: 49395819-49395819 on Build GRCh38
3835371 c.359C>T p.T120I Chr.19: 49395792-49395792 on Build GRCh38
3938208 c.352G>T p.E118* Chr.19: 49395785-49395785 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs987577343 Chr.19: 49395830-49395830 on Build GRCh38
rs1055944238 Chr.19: 49395946-49395946 on Build GRCh38
rs887028561 Chr.19: 49395889-49395889 on Build GRCh38
rs1046930096 Chr.19: 49395880-49395880 on Build GRCh38
rs115156590 Chr.19: 49395872-49395872 on Build GRCh38
rs8101815 Chr.19: 49395947-49395947 on Build GRCh38
rs962272640 Chr.19: 49395864-49395864 on Build GRCh38
rs751740217 Chr.19: 49395810-49395810 on Build GRCh38
rs766540572 Chr.19: 49395806-49395806 on Build GRCh38
rs10405154 Chr.19: 49395819-49395819 on Build GRCh38
rs907920588 Chr.19: 49395841-49395841 on Build GRCh38
rs200504503 Chr.19: 49395797-49395797 on Build GRCh38
rs559172057 Chr.19: 49395818-49395818 on Build GRCh38
rs1014017883 Chr.19: 49395769-49395769 on Build GRCh38
rs111581827 Chr.19: 49395917-49395917 on Build GRCh38
rs987590941 Chr.19: 49395768-49395768 on Build GRCh38
rs777705158 Chr.19: 49395792-49395792 on Build GRCh38
rs374539084 Chr.19: 49395829-49395829 on Build GRCh38
rs374730675 Chr.19: 49395794-49395794 on Build GRCh38
rs555441998 Chr.19: 49395933-49395933 on Build GRCh38
rs8102582 Chr.19: 49395780-49395780 on Build GRCh38
rs781483131 Chr.19: 49395816-49395816 on Build GRCh38
rs367668535 Chr.19: 49395932-49395932 on Build GRCh38
rs185007630 Chr.19: 49395790-49395790 on Build GRCh38
rs760661890 Chr.19: 49395774-49395774 on Build GRCh38
rs755124537 Chr.19: 49395814-49395814 on Build GRCh38
rs771282639 Chr.19: 49395877-49395877 on Build GRCh38
rs1032961219 Chr.19: 49395796-49395796 on Build GRCh38
rs370579866 Chr.19: 49395793-49395793 on Build GRCh38
rs539237288 Chr.19: 49395840-49395840 on Build GRCh38

View Full Product Details