Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 30785804-30786029 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM188B
Entrez Gene ID
84182
Gene Name
family with sequence similarity 188 member B
Gene Aliases
C7orf67
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30771417-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Gene Symbol
INMT-FAM188B
Entrez Gene ID
100526825
Gene Name
INMT-FAM188B readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.660192
Gene Chromosome Location
Chr.7: 30752135-30892387 on Build GRCh38
Transcripts
NM_032222.2 NR_037598.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
297366 c.506C>T p.P169L Chr.7: 30785835-30785835 on Build GRCh38
3880578 c.545T>C p.I182T Chr.7: 30785874-30785874 on Build GRCh38
5784204 c.614G>C p.G205A Chr.7: 30785943-30785943 on Build GRCh38
3880577 c.476C>T p.P159L Chr.7: 30785805-30785805 on Build GRCh38
4164296 c.485G>T p.S162I Chr.7: 30785814-30785814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373216241 Chr.7: 30785875-30785875 on Build GRCh38
rs781166574 Chr.7: 30785830-30785830 on Build GRCh38
rs764203674 Chr.7: 30785883-30785883 on Build GRCh38
rs771886071 Chr.7: 30785874-30785874 on Build GRCh38
rs370755966 Chr.7: 30785932-30785932 on Build GRCh38
rs750196986 Chr.7: 30785834-30785834 on Build GRCh38
rs979093415 Chr.7: 30785937-30785937 on Build GRCh38
rs75843887 Chr.7: 30785928-30785928 on Build GRCh38
rs567710671 Chr.7: 30785957-30785957 on Build GRCh38
rs762119316 Chr.7: 30785903-30785903 on Build GRCh38
rs770112487 Chr.7: 30785960-30785960 on Build GRCh38
rs111491161 Chr.7: 30785835-30785835 on Build GRCh38
rs775962382 Chr.7: 30785881-30785881 on Build GRCh38
rs1019366557 Chr.7: 30785997-30785997 on Build GRCh38
rs372001411 Chr.7: 30785993-30785993 on Build GRCh38
rs774490751 Chr.7: 30785983-30785983 on Build GRCh38
rs73685795 Chr.7: 30785987-30785987 on Build GRCh38
rs769002806 Chr.7: 30785978-30785978 on Build GRCh38
rs965106733 Chr.7: 30785998-30785998 on Build GRCh38
rs745560380 Chr.7: 30785852-30785852 on Build GRCh38
rs775910067 Chr.7: 30785975-30785975 on Build GRCh38
rs185974904 Chr.7: 30785977-30785977 on Build GRCh38
rs770351539 Chr.7: 30785880-30785880 on Build GRCh38
rs751792153 Chr.7: 30785874-30785875 on Build GRCh38
rs138639794 Chr.7: 30785889-30785889 on Build GRCh38
rs768186750 Chr.7: 30785838-30785838 on Build GRCh38
rs530211573 Chr.7: 30785929-30785929 on Build GRCh38
rs34357272 Chr.7: 30785877-30785877 on Build GRCh38
rs753795286 Chr.7: 30785949-30785949 on Build GRCh38
rs77067592 Chr.7: 30785955-30785955 on Build GRCh38
rs12672184 Chr.7: 30785959-30785959 on Build GRCh38
rs74821332 Chr.7: 30785890-30785890 on Build GRCh38
rs757043983 Chr.7: 30785829-30785829 on Build GRCh38
rs377460893 Chr.7: 30785878-30785878 on Build GRCh38
rs773559665 Chr.7: 30786001-30786001 on Build GRCh38
rs748820864 Chr.7: 30785836-30785836 on Build GRCh38
rs754826617 Chr.7: 30785950-30785950 on Build GRCh38
rs144509099 Chr.7: 30785913-30785913 on Build GRCh38
rs375313491 Chr.7: 30785954-30785954 on Build GRCh38

View Full Product Details