Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 16552821-16553082 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
-
UniGene
Hs.467769
Gene Chromosome Location
Chr.2: 16549459-16665866 on Build GRCh38
Transcripts
NM_030797.3 XM_005262630.3 XM_006712108.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4846443 c.955C>G p.R319G Chr.2: 16552953-16552953 on Build GRCh38
1400711 c.970T>C p.*324Q Chr.2: 16552938-16552938 on Build GRCh38
4999023 c.916A>G p.T306A Chr.2: 16552992-16552992 on Build GRCh38
3569367 c.912C>T p.F304F Chr.2: 16552996-16552996 on Build GRCh38
3569366 c.931G>A p.D311N Chr.2: 16552977-16552977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs999374301 Chr.2: 16553058-16553058 on Build GRCh38
rs368577157 Chr.2: 16552899-16552899 on Build GRCh38
rs377597411 Chr.2: 16553034-16553034 on Build GRCh38
rs755676885 Chr.2: 16552893-16552893 on Build GRCh38
rs568794036 Chr.2: 16552868-16552868 on Build GRCh38
rs778434434 Chr.2: 16553026-16553026 on Build GRCh38
rs373188470 Chr.2: 16553008-16553008 on Build GRCh38
rs747685126 Chr.2: 16552889-16552889 on Build GRCh38
rs1043506322 Chr.2: 16553043-16553043 on Build GRCh38
rs375076153 Chr.2: 16553052-16553052 on Build GRCh38
rs777223445 Chr.2: 16553010-16553010 on Build GRCh38
rs13404748 Chr.2: 16552885-16552885 on Build GRCh38
rs146795138 Chr.2: 16552981-16552981 on Build GRCh38
rs201324134 Chr.2: 16552933-16552933 on Build GRCh38
rs751897701 Chr.2: 16552970-16552970 on Build GRCh38
rs955933629 Chr.2: 16552945-16552945 on Build GRCh38
rs765457820 Chr.2: 16552929-16552929 on Build GRCh38
rs114377299 Chr.2: 16553027-16553027 on Build GRCh38
rs187539517 Chr.2: 16552925-16552925 on Build GRCh38
rs756844443 Chr.2: 16553019-16553019 on Build GRCh38
rs780019685 Chr.2: 16553030-16553030 on Build GRCh38
rs924383055 Chr.2: 16552862-16552862 on Build GRCh38
rs369134296 Chr.2: 16552930-16552930 on Build GRCh38
rs570496174 Chr.2: 16552897-16552897 on Build GRCh38
rs375357516 Chr.2: 16552968-16552968 on Build GRCh38
rs949463983 Chr.2: 16553040-16553040 on Build GRCh38
rs371762080 Chr.2: 16552939-16552939 on Build GRCh38
rs774146267 Chr.2: 16552913-16552913 on Build GRCh38
rs781128503 Chr.2: 16553037-16553038 on Build GRCh38
rs12997119 Chr.2: 16553016-16553016 on Build GRCh38
rs376587125 Chr.2: 16552927-16552927 on Build GRCh38
rs903792850 Chr.2: 16553049-16553049 on Build GRCh38
rs770599023 Chr.2: 16552911-16552911 on Build GRCh38
rs763229345 Chr.2: 16552952-16552952 on Build GRCh38
rs376429680 Chr.2: 16553029-16553029 on Build GRCh38
rs746985999 Chr.2: 16553047-16553047 on Build GRCh38
rs372607588 Chr.2: 16552920-16552920 on Build GRCh38
rs559615335 Chr.2: 16552854-16552854 on Build GRCh38
rs369382237 Chr.2: 16552978-16552978 on Build GRCh38

View Full Product Details