Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156294352-156294597 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLMP
Entrez Gene ID
112770
Gene Name
glycosylated lysosomal membrane protein
Gene Aliases
C1orf85, NCU-G1
UniGene
Hs.202522
Gene Chromosome Location
Chr.1: 156292687-156295689 on Build GRCh38
Transcripts
NM_001256604.1 NM_001256605.1 NM_001256608.1 NM_001256609.1 NM_144580.2 XM_011509117.2 XM_017000149.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3710336 c.288C>G p.T96T Chr.1: 156294398-156294398 on Build GRCh38
897180 c.460G>A p.D154N Chr.1: 156294484-156294484 on Build GRCh38
3475202 c.265G>C p.G89R Chr.1: 156294421-156294421 on Build GRCh38
4023242 c.289A>G p.R97G Chr.1: 156294397-156294397 on Build GRCh38
6577045 c.271C>A p.P91T Chr.1: 156294415-156294415 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs572915309 Chr.1: 156294494-156294494 on Build GRCh38
rs148759491 Chr.1: 156294548-156294548 on Build GRCh38
rs191092436 Chr.1: 156294454-156294454 on Build GRCh38
rs763721532 Chr.1: 156294441-156294441 on Build GRCh38
rs758249381 Chr.1: 156294510-156294510 on Build GRCh38
rs781086840 Chr.1: 156294380-156294380 on Build GRCh38
rs571649198 Chr.1: 156294530-156294530 on Build GRCh38
rs567266121 Chr.1: 156294520-156294520 on Build GRCh38
rs371084985 Chr.1: 156294535-156294535 on Build GRCh38
rs140018765 Chr.1: 156294531-156294531 on Build GRCh38
rs761251022 Chr.1: 156294428-156294428 on Build GRCh38
rs147299774 Chr.1: 156294542-156294542 on Build GRCh38
rs535277966 Chr.1: 156294439-156294439 on Build GRCh38
rs778155687 Chr.1: 156294525-156294525 on Build GRCh38
rs567172789 Chr.1: 156294406-156294406 on Build GRCh38
rs895392812 Chr.1: 156294432-156294432 on Build GRCh38
rs377594661 Chr.1: 156294418-156294418 on Build GRCh38
rs771660426 Chr.1: 156294414-156294414 on Build GRCh38
rs981097686 Chr.1: 156294485-156294485 on Build GRCh38
rs34655212 Chr.1: 156294384-156294384 on Build GRCh38
rs939462393 Chr.1: 156294420-156294420 on Build GRCh38
rs781351658 Chr.1: 156294456-156294458 on Build GRCh38
rs746311147 Chr.1: 156294410-156294410 on Build GRCh38
rs915337700 Chr.1: 156294377-156294377 on Build GRCh38
rs201165533 Chr.1: 156294411-156294411 on Build GRCh38
rs149357476 Chr.1: 156294484-156294484 on Build GRCh38
rs374408285 Chr.1: 156294536-156294536 on Build GRCh38
rs767677309 Chr.1: 156294513-156294513 on Build GRCh38
rs752716142 Chr.1: 156294517-156294517 on Build GRCh38
rs772855023 Chr.1: 156294577-156294577 on Build GRCh38
rs769630354 Chr.1: 156294567-156294567 on Build GRCh38
rs771364866 Chr.1: 156294407-156294407 on Build GRCh38
rs756375095 Chr.1: 156294389-156294389 on Build GRCh38
rs368809546 Chr.1: 156294575-156294575 on Build GRCh38

View Full Product Details