Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67828456-67828711 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPT
Entrez Gene ID
80152
Gene Name
centromere protein T
Gene Aliases
C16orf56, CENP-T
UniGene
Hs.288382
Gene Chromosome Location
Chr.16: 67828157-67847458 on Build GRCh38
Transcripts
NM_025082.3 XM_006721277.2 XM_011523344.1 XM_011523345.1 XM_017023712.1 XM_017023713.1 XM_017023714.1 XM_017023715.1 XM_017023716.1 XM_017023717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6229017 c.1559G>A p.R520Q Chr.16: 67828477-67828477 on Build GRCh38
5743761 c.1562G>A p.R521Q Chr.16: 67828474-67828474 on Build GRCh38
7415735 c.1532A>G p.K511R Chr.16: 67828504-67828504 on Build GRCh38
7165941 c.1524G>A p.K508K Chr.16: 67828512-67828512 on Build GRCh38
1679190 c.1557G>A p.M519I Chr.16: 67828479-67828479 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755650297 Chr.16: 67828649-67828649 on Build GRCh38
rs749623085 Chr.16: 67828550-67828550 on Build GRCh38
rs371458502 Chr.16: 67828605-67828605 on Build GRCh38
rs372241354 Chr.16: 67828660-67828660 on Build GRCh38
rs777661469 Chr.16: 67828492-67828492 on Build GRCh38
rs866534418 Chr.16: 67828664-67828666 on Build GRCh38
rs933603122 Chr.16: 67828497-67828497 on Build GRCh38
rs749748044 Chr.16: 67828655-67828655 on Build GRCh38
rs758161409 Chr.16: 67828490-67828490 on Build GRCh38
rs200300063 Chr.16: 67828477-67828477 on Build GRCh38
rs376562503 Chr.16: 67828627-67828627 on Build GRCh38
rs779590867 Chr.16: 67828654-67828654 on Build GRCh38
rs1035757868 Chr.16: 67828652-67828652 on Build GRCh38
rs780558609 Chr.16: 67828537-67828537 on Build GRCh38
rs543219989 Chr.16: 67828474-67828474 on Build GRCh38
rs748560151 Chr.16: 67828664-67828664 on Build GRCh38
rs368630646 Chr.16: 67828657-67828657 on Build GRCh38
rs773600523 Chr.16: 67828683-67828683 on Build GRCh38
rs559580852 Chr.16: 67828528-67828528 on Build GRCh38
rs760321411 Chr.16: 67828689-67828689 on Build GRCh38
rs752652513 Chr.16: 67828615-67828615 on Build GRCh38
rs756991831 Chr.16: 67828507-67828507 on Build GRCh38
rs772347517 Chr.16: 67828560-67828560 on Build GRCh38
rs1053879293 Chr.16: 67828522-67828522 on Build GRCh38
rs780675427 Chr.16: 67828511-67828511 on Build GRCh38
rs927407557 Chr.16: 67828601-67828601 on Build GRCh38
rs563642592 Chr.16: 67828554-67828554 on Build GRCh38
rs767485698 Chr.16: 67828476-67828476 on Build GRCh38
rs760861147 Chr.16: 67828599-67828599 on Build GRCh38
rs202153240 Chr.16: 67828687-67828687 on Build GRCh38
rs752493054 Chr.16: 67828482-67828482 on Build GRCh38
rs530979236 Chr.16: 67828628-67828628 on Build GRCh38
rs751149397 Chr.16: 67828503-67828503 on Build GRCh38
rs117015245 Chr.16: 67828517-67828517 on Build GRCh38
rs1002833089 Chr.16: 67828645-67828645 on Build GRCh38
rs1008047941 Chr.16: 67828520-67828520 on Build GRCh38
rs762994654 Chr.16: 67828625-67828625 on Build GRCh38
rs945288768 Chr.16: 67828651-67828651 on Build GRCh38
rs761940448 Chr.16: 67828558-67828558 on Build GRCh38
rs760512144 Chr.16: 67828584-67828586 on Build GRCh38
rs370858297 Chr.16: 67828475-67828475 on Build GRCh38
rs765280258 Chr.16: 67828479-67828479 on Build GRCh38
rs573619904 Chr.16: 67828516-67828516 on Build GRCh38
rs1011428486 Chr.16: 67828612-67828612 on Build GRCh38
rs545377044 Chr.16: 67828552-67828552 on Build GRCh38
rs533198810 Chr.16: 67828529-67828529 on Build GRCh38
rs772372421 Chr.16: 67828676-67828676 on Build GRCh38
rs780832457 Chr.16: 67828632-67828632 on Build GRCh38
rs773369180 Chr.16: 67828573-67828573 on Build GRCh38
rs759480785 Chr.16: 67828478-67828478 on Build GRCh38

View Full Product Details