Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 181715949-181716218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACNA1E
Entrez Gene ID
777
Gene Name
calcium voltage-gated channel subunit alpha1 E
Gene Aliases
BII, CACH6, CACNL1A6, Cav2.3, gm139
UniGene
Hs.437444
Gene Chromosome Location
Chr.1: 181317712-181808084 on Build GRCh38
Transcripts
NM_000721.3 NM_001205293.1 NM_001205294.1 XM_017002243.1 XM_017002244.1 XM_017002245.1 XM_017002246.1 XM_017002247.1 XM_017002248.1 XM_017002249.1 XM_017002250.1 XM_017002251.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6541378 c.71G>T p.R24M Chr.1: 181716064-181716064 on Build GRCh38
6541375 c.1250G>T p.R417M Chr.1: 181716064-181716064 on Build GRCh38
5207187 c.1159T>C p.S387P Chr.1: 181716120-181716120 on Build GRCh38
4226875 c.71G>A p.R24K Chr.1: 181716064-181716064 on Build GRCh38
1473106 c.1305C>A p.I435I Chr.1: 181716119-181716119 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1014332101 Chr.1: 181716019-181716019 on Build GRCh38
rs377231001 Chr.1: 181716069-181716069 on Build GRCh38
rs938549648 Chr.1: 181716073-181716073 on Build GRCh38
rs575438278 Chr.1: 181716111-181716111 on Build GRCh38
rs937242865 Chr.1: 181715985-181715985 on Build GRCh38
rs758710221 Chr.1: 181716018-181716018 on Build GRCh38
rs567371921 Chr.1: 181716179-181716179 on Build GRCh38
rs533180173 Chr.1: 181716017-181716017 on Build GRCh38
rs925889513 Chr.1: 181715984-181715984 on Build GRCh38
rs768993953 Chr.1: 181716126-181716126 on Build GRCh38
rs548919740 Chr.1: 181716173-181716173 on Build GRCh38
rs924424984 Chr.1: 181716054-181716054 on Build GRCh38
rs898846338 Chr.1: 181716159-181716159 on Build GRCh38
rs1036148850 Chr.1: 181716089-181716089 on Build GRCh38
rs746405819 Chr.1: 181716081-181716081 on Build GRCh38
rs1057419158 Chr.1: 181716079-181716079 on Build GRCh38
rs757401430 Chr.1: 181716051-181716051 on Build GRCh38
rs374615606 Chr.1: 181716015-181716015 on Build GRCh38
rs866217418 Chr.1: 181716186-181716186 on Build GRCh38
rs772914867 Chr.1: 181716133-181716133 on Build GRCh38
rs559265111 Chr.1: 181716009-181716009 on Build GRCh38
rs747233333 Chr.1: 181716036-181716036 on Build GRCh38
rs1027133730 Chr.1: 181716187-181716187 on Build GRCh38
rs945438203 Chr.1: 181716001-181716001 on Build GRCh38
rs114088252 Chr.1: 181716122-181716122 on Build GRCh38
rs868244118 Chr.1: 181716129-181716129 on Build GRCh38
rs559817705 Chr.1: 181716095-181716095 on Build GRCh38
rs78231578 Chr.1: 181716102-181716102 on Build GRCh38
rs551308908 Chr.1: 181716088-181716088 on Build GRCh38
rs993640969 Chr.1: 181716171-181716171 on Build GRCh38
rs866035995 Chr.1: 181716070-181716070 on Build GRCh38
rs778202285 Chr.1: 181716035-181716035 on Build GRCh38
rs765937003 Chr.1: 181716161-181716161 on Build GRCh38
rs373180436 Chr.1: 181716141-181716141 on Build GRCh38
rs866705227 Chr.1: 181716121-181716121 on Build GRCh38

View Full Product Details