Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31641183-31641442 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAG6
Entrez Gene ID
7917
Gene Name
BCL2 associated athanogene 6
Gene Aliases
BAG-6, BAT3, D6S52E, G3
UniGene
Hs.440900
Gene Chromosome Location
Chr.6: 31639028-31660900 on Build GRCh38
Transcripts
NM_001098534.1 NM_001199697.1 NM_001199698.1 NM_004639.3 NM_080702.2 NM_080703.2 XM_011514892.2 XM_017011275.1 XM_017011276.1 XM_017011277.1 XM_017011278.1 XM_017011279.1 XM_017011280.1 XM_017011281.1 XM_017011282.1 XM_017011283.1 XM_017011284.1 XM_017011285.1 XM_017011286.1 XM_017011287.1 XM_017011288.1 XM_017011289.1 XM_017011290.1 XM_017011291.1 XM_017011292.1 XM_017011293.1 XM_017011294.1 XM_017011295.1 XM_017011296.1 XM_017011297.1 XM_017011298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7202178 c.2474G>A p.G825D Chr.6: 31641400-31641400 on Build GRCh38
6173646 c.2592C>T p.I864I Chr.6: 31641390-31641390 on Build GRCh38
6875813 c.2616C>G p.L872L Chr.6: 31641366-31641366 on Build GRCh38
3253670 c.2568G>A p.V856V Chr.6: 31641414-31641414 on Build GRCh38
6992659 c.2603A>T p.N868I Chr.6: 31641379-31641379 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771679482 Chr.6: 31641315-31641316 on Build GRCh38
rs142878083 Chr.6: 31641339-31641339 on Build GRCh38
rs768945156 Chr.6: 31641324-31641324 on Build GRCh38
rs761958293 Chr.6: 31641336-31641336 on Build GRCh38
rs150629776 Chr.6: 31641385-31641385 on Build GRCh38
rs4624899 Chr.6: 31641292-31641292 on Build GRCh38
rs571201831 Chr.6: 31641265-31641265 on Build GRCh38
rs757662570 Chr.6: 31641281-31641281 on Build GRCh38
rs753564629 Chr.6: 31641210-31641210 on Build GRCh38
rs775849384 Chr.6: 31641311-31641311 on Build GRCh38
rs374720969 Chr.6: 31641312-31641312 on Build GRCh38
rs781765003 Chr.6: 31641288-31641288 on Build GRCh38
rs376286216 Chr.6: 31641263-31641263 on Build GRCh38
rs535174069 Chr.6: 31641301-31641301 on Build GRCh38
rs764858905 Chr.6: 31641248-31641248 on Build GRCh38
rs752416891 Chr.6: 31641260-31641260 on Build GRCh38
rs980215599 Chr.6: 31641255-31641255 on Build GRCh38
rs546902137 Chr.6: 31641302-31641302 on Build GRCh38
rs927281909 Chr.6: 31641290-31641290 on Build GRCh38
rs201252510 Chr.6: 31641330-31641330 on Build GRCh38
rs369107576 Chr.6: 31641373-31641373 on Build GRCh38
rs746547137 Chr.6: 31641279-31641279 on Build GRCh38
rs754783298 Chr.6: 31641242-31641242 on Build GRCh38
rs1051189181 Chr.6: 31641366-31641366 on Build GRCh38
rs561759974 Chr.6: 31641387-31641387 on Build GRCh38
rs139014684 Chr.6: 31641389-31641389 on Build GRCh38
rs776609828 Chr.6: 31641340-31641340 on Build GRCh38

View Full Product Details