Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59812596-59812841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC6
Entrez Gene ID
8030
Gene Name
coiled-coil domain containing 6
Gene Aliases
D10S170, H4, PTC, TPC, TST1
UniGene
Hs.591360
Gene Chromosome Location
Chr.10: 59788747-59906656 on Build GRCh38
Transcripts
NM_005436.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7430378 c.802G>A p.E268K Chr.10: 59812680-59812680 on Build GRCh38
6456702 c.714C>A p.P238P Chr.10: 59812768-59812768 on Build GRCh38
6562534 c.687-1G>A p.? Chr.10: 59812796-59812796 on Build GRCh38
2148454 c.716T>C p.V239A Chr.10: 59812766-59812766 on Build GRCh38
919202 c.843A>G p.L281L Chr.10: 59812639-59812639 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749668592 Chr.10: 59812755-59812755 on Build GRCh38
rs775541001 Chr.10: 59812767-59812767 on Build GRCh38
rs780829046 Chr.10: 59812670-59812670 on Build GRCh38
rs369677538 Chr.10: 59812812-59812812 on Build GRCh38
rs760941928 Chr.10: 59812768-59812768 on Build GRCh38
rs868314380 Chr.10: 59812760-59812760 on Build GRCh38
rs774944149 Chr.10: 59812765-59812765 on Build GRCh38
rs1043914419 Chr.10: 59812660-59812660 on Build GRCh38
rs753943792 Chr.10: 59812808-59812808 on Build GRCh38
rs780048431 Chr.10: 59812623-59812623 on Build GRCh38
rs762841140 Chr.10: 59812811-59812811 on Build GRCh38
rs771159444 Chr.10: 59812759-59812759 on Build GRCh38
rs1028324450 Chr.10: 59812806-59812806 on Build GRCh38
rs751415417 Chr.10: 59812631-59812631 on Build GRCh38
rs376774410 Chr.10: 59812771-59812771 on Build GRCh38
rs141265324 Chr.10: 59812766-59812766 on Build GRCh38
rs189149234 Chr.10: 59812750-59812750 on Build GRCh38
rs374316271 Chr.10: 59812645-59812645 on Build GRCh38
rs778058230 Chr.10: 59812753-59812753 on Build GRCh38

View Full Product Details