Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 154330753-154331006 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP8B2
Entrez Gene ID
57198
Gene Name
ATPase phospholipid transporting 8B2
Gene Aliases
ATPID
UniGene
Hs.435700
Gene Chromosome Location
Chr.1: 154325541-154351307 on Build GRCh38
Transcripts
NM_001005855.1 NM_020452.3 XM_005245355.2 XM_005245356.3 XM_017001871.1 XM_017001872.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
222565 c.303+1G>A p.? Chr.1: 154330929-154330929 on Build GRCh38
6882734 c.128C>T p.T43I Chr.1: 154330852-154330852 on Build GRCh38
6035391 c.243C>T p.N81N Chr.1: 154330868-154330868 on Build GRCh38
6035393 c.201C>T p.N67N Chr.1: 154330868-154330868 on Build GRCh38
1472690 c.223C>G p.L75V Chr.1: 154330848-154330848 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778845759 Chr.1: 154330899-154330899 on Build GRCh38
rs776254470 Chr.1: 154330829-154330829 on Build GRCh38
rs146860968 Chr.1: 154330869-154330869 on Build GRCh38
rs768331419 Chr.1: 154330824-154330824 on Build GRCh38
rs991494107 Chr.1: 154330960-154330960 on Build GRCh38
rs771629590 Chr.1: 154330805-154330805 on Build GRCh38
rs767649781 Chr.1: 154330871-154330871 on Build GRCh38
rs762480331 Chr.1: 154330834-154330834 on Build GRCh38
rs768423229 Chr.1: 154330946-154330946 on Build GRCh38
rs890289113 Chr.1: 154330922-154330922 on Build GRCh38
rs141275731 Chr.1: 154330822-154330822 on Build GRCh38
rs1037437541 Chr.1: 154330807-154330807 on Build GRCh38
rs773451517 Chr.1: 154330853-154330853 on Build GRCh38
rs1033048374 Chr.1: 154330942-154330942 on Build GRCh38
rs762881967 Chr.1: 154330948-154330948 on Build GRCh38
rs374119776 Chr.1: 154330800-154330800 on Build GRCh38
rs758164152 Chr.1: 154330910-154330910 on Build GRCh38
rs148490903 Chr.1: 154330868-154330868 on Build GRCh38
rs745682103 Chr.1: 154330908-154330908 on Build GRCh38
rs752758454 Chr.1: 154330880-154330880 on Build GRCh38
rs766257787 Chr.1: 154330954-154330954 on Build GRCh38
rs200148570 Chr.1: 154330974-154330974 on Build GRCh38
rs757181484 Chr.1: 154330793-154330793 on Build GRCh38
rs372538054 Chr.1: 154330941-154330941 on Build GRCh38
rs766528266 Chr.1: 154330867-154330867 on Build GRCh38
rs772857544 Chr.1: 154330797-154330798 on Build GRCh38
rs554905339 Chr.1: 154330837-154330837 on Build GRCh38
rs1020940099 Chr.1: 154330889-154330889 on Build GRCh38
rs763333101 Chr.1: 154330860-154330860 on Build GRCh38
rs565503703 Chr.1: 154330937-154330937 on Build GRCh38
rs747607166 Chr.1: 154330968-154330968 on Build GRCh38
rs769072068 Chr.1: 154330811-154330811 on Build GRCh38
rs757197837 Chr.1: 154330890-154330890 on Build GRCh38
rs373800163 Chr.1: 154330780-154330780 on Build GRCh38
rs375613302 Chr.1: 154330958-154330958 on Build GRCh38

View Full Product Details