Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 8365855-8366096 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL4
Entrez Gene ID
51129
Gene Name
angiopoietin like 4
Gene Aliases
ARP4, FIAF, HARP, HFARP, NL2, PGAR, TGQTL, UNQ171, pp1158
UniGene
Hs.9613
Gene Chromosome Location
Chr.19: 8364127-8374375 on Build GRCh38
Transcripts
NM_001039667.2 NM_139314.2 NR_104213.1 XM_005272484.3 XM_005272485.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
227632 c.407G>A p.R136Q Chr.19: 8366042-8366042 on Build GRCh38
3836194 c.327C>G p.L109L Chr.19: 8365962-8365962 on Build GRCh38
3541586 c.393G>A p.E131E Chr.19: 8366028-8366028 on Build GRCh38
3681119 c.429G>A p.Q143Q Chr.19: 8366064-8366064 on Build GRCh38
7262390 c.375G>A p.Q125Q Chr.19: 8366010-8366010 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs577445388 Chr.19: 8365951-8365951 on Build GRCh38
rs374033592 Chr.19: 8365946-8365946 on Build GRCh38
rs749378028 Chr.19: 8366018-8366018 on Build GRCh38
rs376394055 Chr.19: 8365971-8365971 on Build GRCh38
rs773172996 Chr.19: 8365933-8365933 on Build GRCh38
rs1034870122 Chr.19: 8365879-8365879 on Build GRCh38
rs370940440 Chr.19: 8365931-8365931 on Build GRCh38
rs756775279 Chr.19: 8365979-8365979 on Build GRCh38
rs750042693 Chr.19: 8365995-8365995 on Build GRCh38
rs376327761 Chr.19: 8365918-8365918 on Build GRCh38
rs777171576 Chr.19: 8366074-8366074 on Build GRCh38
rs761098321 Chr.19: 8365960-8365960 on Build GRCh38
rs768024878 Chr.19: 8365957-8365957 on Build GRCh38
rs768583490 Chr.19: 8365932-8365932 on Build GRCh38
rs774532632 Chr.19: 8366033-8366033 on Build GRCh38
rs778475399 Chr.19: 8365986-8365986 on Build GRCh38
rs758001199 Chr.19: 8365997-8365997 on Build GRCh38
rs557133449 Chr.19: 8366042-8366042 on Build GRCh38
rs768545199 Chr.19: 8366003-8366003 on Build GRCh38
rs779840915 Chr.19: 8365998-8365998 on Build GRCh38
rs150015993 Chr.19: 8365927-8365927 on Build GRCh38
rs11544610 Chr.19: 8366002-8366002 on Build GRCh38
rs771118296 Chr.19: 8365943-8365943 on Build GRCh38
rs756972923 Chr.19: 8365919-8365925 on Build GRCh38
rs1011478238 Chr.19: 8366019-8366033 on Build GRCh38
rs753061344 Chr.19: 8365959-8365959 on Build GRCh38
rs781178060 Chr.19: 8366017-8366017 on Build GRCh38
rs181710838 Chr.19: 8366075-8366075 on Build GRCh38
rs771829106 Chr.19: 8365909-8365909 on Build GRCh38
rs771012831 Chr.19: 8366028-8366028 on Build GRCh38
rs373549197 Chr.19: 8365935-8365935 on Build GRCh38
rs774693784 Chr.19: 8365945-8365945 on Build GRCh38
rs776000614 Chr.19: 8366037-8366037 on Build GRCh38
rs753353884 Chr.19: 8365974-8365974 on Build GRCh38
rs746114842 Chr.19: 8366034-8366034 on Build GRCh38
rs764563916 Chr.19: 8366050-8366050 on Build GRCh38
rs373652079 Chr.19: 8365916-8365916 on Build GRCh38

View Full Product Details