Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 87901743-87902016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5A
Entrez Gene ID
763
Gene Name
carbonic anhydrase 5A
Gene Aliases
CA5, CA5AD, CAV, CAVA, GS1-21A4.1
UniGene
Hs.177446
Gene Chromosome Location
Chr.16: 87888019-87936575 on Build GRCh38
Transcripts
NM_001739.1 XM_011523309.2 XM_011523310.2 XM_017023646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6195247 c.594C>A p.D198E Chr.16: 87901936-87901936 on Build GRCh38
6080301 c.585G>T p.R195S Chr.16: 87901945-87901945 on Build GRCh38
1578584 c.608T>C p.I203T Chr.16: 87901922-87901922 on Build GRCh38
6617120 c.601C>T p.P201S Chr.16: 87901929-87901929 on Build GRCh38
6022743 c.583A>G p.R195G Chr.16: 87901947-87901947 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776492248 Chr.16: 87901934-87901934 on Build GRCh38
rs780315322 Chr.16: 87901907-87901907 on Build GRCh38
rs557809782 Chr.16: 87901775-87901775 on Build GRCh38
rs576285826 Chr.16: 87901776-87901776 on Build GRCh38
rs764657276 Chr.16: 87901885-87901885 on Build GRCh38
rs923718683 Chr.16: 87901788-87901788 on Build GRCh38
rs745458044 Chr.16: 87901991-87901991 on Build GRCh38
rs139856895 Chr.16: 87901778-87901778 on Build GRCh38
rs769184030 Chr.16: 87901834-87901834 on Build GRCh38
rs754005998 Chr.16: 87901954-87901954 on Build GRCh38
rs762183629 Chr.16: 87901937-87901937 on Build GRCh38
rs372374928 Chr.16: 87901972-87901972 on Build GRCh38
rs139816901 Chr.16: 87901915-87901915 on Build GRCh38
rs750775991 Chr.16: 87901938-87901938 on Build GRCh38
rs767721061 Chr.16: 87901893-87901893 on Build GRCh38
rs145215237 Chr.16: 87901955-87901955 on Build GRCh38
rs770692285 Chr.16: 87901933-87901933 on Build GRCh38
rs938984743 Chr.16: 87901798-87901798 on Build GRCh38
rs367977671 Chr.16: 87901882-87901882 on Build GRCh38
rs761097356 Chr.16: 87901940-87901940 on Build GRCh38
rs756294988 Chr.16: 87901902-87901902 on Build GRCh38
rs758247579 Chr.16: 87901969-87901969 on Build GRCh38
rs762391118 Chr.16: 87901892-87901892 on Build GRCh38
rs774224687 Chr.16: 87901872-87901872 on Build GRCh38
rs1056162584 Chr.16: 87901817-87901817 on Build GRCh38
rs760364703 Chr.16: 87901879-87901879 on Build GRCh38
rs1049457872 Chr.16: 87901860-87901860 on Build GRCh38
rs751943757 Chr.16: 87901886-87901886 on Build GRCh38
rs373576729 Chr.16: 87901878-87901878 on Build GRCh38
rs370038828 Chr.16: 87901815-87901815 on Build GRCh38
rs555610444 Chr.16: 87901807-87901807 on Build GRCh38
rs780065663 Chr.16: 87901866-87901866 on Build GRCh38
rs772079493 Chr.16: 87901930-87901930 on Build GRCh38
rs997575240 Chr.16: 87901904-87901904 on Build GRCh38
rs894829795 Chr.16: 87901822-87901822 on Build GRCh38
rs778818832 Chr.16: 87901967-87901967 on Build GRCh38
rs769123644 Chr.16: 87901935-87901935 on Build GRCh38
rs990325883 Chr.16: 87901944-87901944 on Build GRCh38
rs777682092 Chr.16: 87901932-87901932 on Build GRCh38
rs779068362 Chr.16: 87901917-87901917 on Build GRCh38
rs374788491 Chr.16: 87901883-87901883 on Build GRCh38
rs372389355 Chr.16: 87901987-87901987 on Build GRCh38
rs372043258 Chr.16: 87901903-87901903 on Build GRCh38
rs750814782 Chr.16: 87901898-87901898 on Build GRCh38
rs746028026 Chr.16: 87901863-87901863 on Build GRCh38
rs373781815 Chr.16: 87901877-87901877 on Build GRCh38
rs533205037 Chr.16: 87901867-87901867 on Build GRCh38
rs189523393 Chr.16: 87901927-87901927 on Build GRCh38
rs775083534 Chr.16: 87901936-87901936 on Build GRCh38
rs545376802 Chr.16: 87901868-87901868 on Build GRCh38
rs753913199 Chr.16: 87901914-87901914 on Build GRCh38
rs185157091 Chr.16: 87901864-87901864 on Build GRCh38
rs369715098 Chr.16: 87901941-87901941 on Build GRCh38
rs375321548 Chr.16: 87901974-87901974 on Build GRCh38
rs769513594 Chr.16: 87901995-87901995 on Build GRCh38
rs568286213 Chr.16: 87901856-87901856 on Build GRCh38
rs752863808 Chr.16: 87901968-87901968 on Build GRCh38
rs376698350 Chr.16: 87901814-87901814 on Build GRCh38

View Full Product Details