Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 113767212-113767437 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHS1
Entrez Gene ID
79949
Gene Name
pleckstrin homology domain containing S1
Gene Aliases
C10orf81, HEL185, bA211N11.2
UniGene
Hs.228320
Gene Chromosome Location
Chr.10: 113751262-113782433 on Build GRCh38
Transcripts
NM_001193434.1 NM_001193435.1 NM_024889.4 NM_182601.1 XM_005270162.2 XM_005270163.2 XM_011540171.1 XM_011540172.1 XM_011540173.1 XM_011540174.2 XM_011540175.1 XM_011540176.1 XM_017016666.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4404725 c.263A>C p.K88T Chr.10: 113767383-113767383 on Build GRCh38
1128135 c.309C>T p.V103V Chr.10: 113767429-113767429 on Build GRCh38
5453700 c.255C>T p.S85S Chr.10: 113767375-113767375 on Build GRCh38
1128134 c.45C>T p.V15V Chr.10: 113767429-113767429 on Build GRCh38
6435979 c.245A>T p.K82M Chr.10: 113767383-113767383 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764105171 Chr.10: 113767312-113767312 on Build GRCh38
rs958138056 Chr.10: 113767270-113767270 on Build GRCh38
rs1046703692 Chr.10: 113767288-113767288 on Build GRCh38
rs756793947 Chr.10: 113767342-113767342 on Build GRCh38
rs546667442 Chr.10: 113767324-113767324 on Build GRCh38
rs749597479 Chr.10: 113767362-113767362 on Build GRCh38
rs755681942 Chr.10: 113767354-113767354 on Build GRCh38
rs773119971 Chr.10: 113767390-113767390 on Build GRCh38
rs921483443 Chr.10: 113767365-113767365 on Build GRCh38
rs73361839 Chr.10: 113767289-113767289 on Build GRCh38
rs748695759 Chr.10: 113767378-113767378 on Build GRCh38
rs199935916 Chr.10: 113767331-113767331 on Build GRCh38
rs779760191 Chr.10: 113767356-113767356 on Build GRCh38
rs779389095 Chr.10: 113767373-113767373 on Build GRCh38
rs1032441161 Chr.10: 113767251-113767251 on Build GRCh38
rs754808731 Chr.10: 113767249-113767249 on Build GRCh38
rs144206508 Chr.10: 113767352-113767352 on Build GRCh38
rs958131264 Chr.10: 113767299-113767299 on Build GRCh38
rs767283570 Chr.10: 113767259-113767259 on Build GRCh38
rs151023522 Chr.10: 113767363-113767363 on Build GRCh38
rs371522471 Chr.10: 113767296-113767296 on Build GRCh38
rs747146672 Chr.10: 113767400-113767400 on Build GRCh38
rs770995901 Chr.10: 113767415-113767415 on Build GRCh38
rs772171711 Chr.10: 113767386-113767386 on Build GRCh38
rs766982824 Chr.10: 113767350-113767350 on Build GRCh38
rs142162556 Chr.10: 113767304-113767304 on Build GRCh38
rs200805404 Chr.10: 113767295-113767295 on Build GRCh38

View Full Product Details