Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 111467543-111467816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTG4
Entrez Gene ID
54766
Gene Name
BTG anti-proliferation factor 4
Gene Aliases
APRO3, PC3B
UniGene
Hs.128180
Gene Chromosome Location
Chr.11: 111383828-111512388 on Build GRCh38
Transcripts
NM_017589.3 XM_011542876.2 XM_011542878.2 XM_011542880.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3415521 c.667T>G p.L223V Chr.11: 111467677-111467677 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs12803406 Chr.11: 111467603-111467603 on Build GRCh38
rs541777980 Chr.11: 111467749-111467749 on Build GRCh38
rs145399036 Chr.11: 111467679-111467679 on Build GRCh38
rs651882 Chr.11: 111467753-111467753 on Build GRCh38
rs770958470 Chr.11: 111467721-111467721 on Build GRCh38
rs760066896 Chr.11: 111467670-111467670 on Build GRCh38
rs762950930 Chr.11: 111467649-111467649 on Build GRCh38
rs778561909 Chr.11: 111467684-111467684 on Build GRCh38
rs774453865 Chr.11: 111467728-111467728 on Build GRCh38
rs139024566 Chr.11: 111467594-111467594 on Build GRCh38
rs878903608 Chr.11: 111467663-111467663 on Build GRCh38
rs529681139 Chr.11: 111467736-111467736 on Build GRCh38
rs768766238 Chr.11: 111467707-111467707 on Build GRCh38
rs757932307 Chr.11: 111467690-111467690 on Build GRCh38
rs779745416 Chr.11: 111467695-111467695 on Build GRCh38
rs746965581 Chr.11: 111467627-111467627 on Build GRCh38
rs191144936 Chr.11: 111467630-111467630 on Build GRCh38
rs751954192 Chr.11: 111467771-111467771 on Build GRCh38
rs767741252 Chr.11: 111467671-111467671 on Build GRCh38
rs376898731 Chr.11: 111467752-111467752 on Build GRCh38
rs768535385 Chr.11: 111467629-111467629 on Build GRCh38
rs746901109 Chr.11: 111467729-111467730 on Build GRCh38
rs750057650 Chr.11: 111467689-111467689 on Build GRCh38
rs753186275 Chr.11: 111467635-111467635 on Build GRCh38
rs74881067 Chr.11: 111467589-111467589 on Build GRCh38
rs559381354 Chr.11: 111467729-111467729 on Build GRCh38
rs773158688 Chr.11: 111467644-111467644 on Build GRCh38
rs562696842 Chr.11: 111467595-111467595 on Build GRCh38
rs935733509 Chr.11: 111467577-111467577 on Build GRCh38
rs11603646 Chr.11: 111467708-111467708 on Build GRCh38
rs1004977599 Chr.11: 111467599-111467599 on Build GRCh38
rs966825903 Chr.11: 111467704-111467704 on Build GRCh38
rs368347511 Chr.11: 111467650-111467650 on Build GRCh38
rs1033583385 Chr.11: 111467641-111467641 on Build GRCh38
rs34804032 Chr.11: 111467717-111467717 on Build GRCh38
rs769477321 Chr.11: 111467715-111467715 on Build GRCh38
rs752012810 Chr.11: 111467656-111467656 on Build GRCh38
rs753152297 Chr.11: 111467675-111467675 on Build GRCh38
rs149916604 Chr.11: 111467718-111467718 on Build GRCh38

View Full Product Details