Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 89790936-89791161 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANPEP
Entrez Gene ID
290
Gene Name
alanyl aminopeptidase, membrane
Gene Aliases
APN, CD13, GP150, LAP1, P150, PEPN
UniGene
Hs.1239
Gene Chromosome Location
Chr.15: 89784895-89814852 on Build GRCh38
Transcripts
NM_001150.2 XM_005254892.4 XM_011521473.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1180823 c.2664delT p.F888fs*18 Chr.15: 89790958-89790958 on Build GRCh38
4128566 c.2613T>C p.I871I Chr.15: 89791009-89791009 on Build GRCh38
5012934 c.2635T>C p.F879L Chr.15: 89790987-89790987 on Build GRCh38
7120163 c.2668G>T p.D890Y Chr.15: 89790954-89790954 on Build GRCh38
5470415 c.2602A>G p.N868D Chr.15: 89791020-89791020 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779764342 Chr.15: 89791098-89791098 on Build GRCh38
rs372938510 Chr.15: 89791079-89791079 on Build GRCh38
rs74766672 Chr.15: 89791078-89791078 on Build GRCh38
rs7168329 Chr.15: 89791114-89791114 on Build GRCh38
rs752797827 Chr.15: 89790984-89790984 on Build GRCh38
rs1027942019 Chr.15: 89791093-89791093 on Build GRCh38
rs781266711 Chr.15: 89790996-89790996 on Build GRCh38
rs752887360 Chr.15: 89791071-89791071 on Build GRCh38
rs776930737 Chr.15: 89791060-89791060 on Build GRCh38
rs773650895 Chr.15: 89791051-89791051 on Build GRCh38
rs768277577 Chr.15: 89791019-89791019 on Build GRCh38
rs200363198 Chr.15: 89791100-89791100 on Build GRCh38
rs771502186 Chr.15: 89791135-89791135 on Build GRCh38
rs781008050 Chr.15: 89791014-89791014 on Build GRCh38
rs148098345 Chr.15: 89791042-89791042 on Build GRCh38
rs973825926 Chr.15: 89791090-89791090 on Build GRCh38
rs369201239 Chr.15: 89791059-89791059 on Build GRCh38
rs765520167 Chr.15: 89791070-89791070 on Build GRCh38
rs757296390 Chr.15: 89790994-89790994 on Build GRCh38
rs144085303 Chr.15: 89790973-89790973 on Build GRCh38
rs1015428579 Chr.15: 89791066-89791066 on Build GRCh38
rs767570451 Chr.15: 89791080-89791080 on Build GRCh38
rs1012837008 Chr.15: 89791018-89791018 on Build GRCh38
rs748800977 Chr.15: 89791131-89791131 on Build GRCh38
rs755962012 Chr.15: 89791097-89791097 on Build GRCh38
rs750290072 Chr.15: 89791089-89791089 on Build GRCh38
rs747422950 Chr.15: 89791138-89791138 on Build GRCh38
rs376323078 Chr.15: 89790972-89790972 on Build GRCh38
rs745692624 Chr.15: 89790999-89790999 on Build GRCh38
rs25654 Chr.15: 89791009-89791009 on Build GRCh38
rs376865285 Chr.15: 89791134-89791134 on Build GRCh38
rs761103812 Chr.15: 89791058-89791058 on Build GRCh38
rs986997722 Chr.15: 89791102-89791102 on Build GRCh38
rs143655390 Chr.15: 89791015-89791015 on Build GRCh38
rs758736666 Chr.15: 89791095-89791096 on Build GRCh38
rs759722593 Chr.15: 89791069-89791069 on Build GRCh38
rs778063366 Chr.15: 89791067-89791067 on Build GRCh38
rs773797848 Chr.15: 89791136-89791136 on Build GRCh38
rs746281500 Chr.15: 89790957-89790958 on Build GRCh38
rs772625359 Chr.15: 89791044-89791044 on Build GRCh38
rs200383502 Chr.15: 89790979-89790979 on Build GRCh38
rs773892132 Chr.15: 89791032-89791032 on Build GRCh38
rs967007676 Chr.15: 89791123-89791123 on Build GRCh38
rs201233331 Chr.15: 89791125-89791125 on Build GRCh38
rs756105308 Chr.15: 89791025-89791025 on Build GRCh38

View Full Product Details