Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 170067216-170067485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AADAT
Entrez Gene ID
51166
Gene Name
aminoadipate aminotransferase
Gene Aliases
KAT2, KATII, KYAT2
UniGene
Hs.529735
Gene Chromosome Location
Chr.4: 170060222-170091699 on Build GRCh38
Transcripts
NM_001286682.1 NM_001286683.1 NM_016228.3 NM_182662.1 XM_006714231.2 XM_011532020.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
447603 c.924C>T p.H308H Chr.4: 170067365-170067365 on Build GRCh38
7099595 c.937G>A p.E313K Chr.4: 170067352-170067352 on Build GRCh38
6589234 c.925G>A p.E309K Chr.4: 170067364-170067364 on Build GRCh38
1309898 c.921A>G p.L307L Chr.4: 170067368-170067368 on Build GRCh38
4657846 c.901-1G>C p.? Chr.4: 170067389-170067389 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs35866167 Chr.4: 170067347-170067348 on Build GRCh38
rs1040254718 Chr.4: 170067266-170067266 on Build GRCh38
rs781071963 Chr.4: 170067330-170067330 on Build GRCh38
rs887441364 Chr.4: 170067316-170067316 on Build GRCh38
rs754742256 Chr.4: 170067428-170067428 on Build GRCh38
rs142229838 Chr.4: 170067378-170067378 on Build GRCh38
rs772938371 Chr.4: 170067268-170067268 on Build GRCh38
rs901747217 Chr.4: 170067277-170067277 on Build GRCh38
rs751338837 Chr.4: 170067321-170067321 on Build GRCh38
rs762346026 Chr.4: 170067395-170067395 on Build GRCh38
rs150671652 Chr.4: 170067317-170067318 on Build GRCh38
rs748131877 Chr.4: 170067361-170067361 on Build GRCh38
rs374213793 Chr.4: 170067283-170067283 on Build GRCh38
rs751248700 Chr.4: 170067427-170067427 on Build GRCh38
rs1053376527 Chr.4: 170067405-170067405 on Build GRCh38
rs758004660 Chr.4: 170067307-170067307 on Build GRCh38
rs777318338 Chr.4: 170067279-170067279 on Build GRCh38
rs972858283 Chr.4: 170067425-170067425 on Build GRCh38
rs372191954 Chr.4: 170067435-170067435 on Build GRCh38
rs749936781 Chr.4: 170067306-170067306 on Build GRCh38
rs574180093 Chr.4: 170067399-170067399 on Build GRCh38
rs368304372 Chr.4: 170067364-170067364 on Build GRCh38
rs562107223 Chr.4: 170067313-170067313 on Build GRCh38
rs183486777 Chr.4: 170067319-170067319 on Build GRCh38
rs777161193 Chr.4: 170067393-170067393 on Build GRCh38
rs754830422 Chr.4: 170067327-170067327 on Build GRCh38
rs779009351 Chr.4: 170067365-170067365 on Build GRCh38
rs764758629 Chr.4: 170067293-170067293 on Build GRCh38
rs747347567 Chr.4: 170067390-170067390 on Build GRCh38
rs769193418 Chr.4: 170067392-170067392 on Build GRCh38
rs1009068004 Chr.4: 170067446-170067447 on Build GRCh38
rs1017810752 Chr.4: 170067451-170067451 on Build GRCh38
rs1028654126 Chr.4: 170067285-170067285 on Build GRCh38
rs763426784 Chr.4: 170067317-170067317 on Build GRCh38
rs772723187 Chr.4: 170067406-170067406 on Build GRCh38
rs992651734 Chr.4: 170067337-170067337 on Build GRCh38
rs368676033 Chr.4: 170067382-170067382 on Build GRCh38
rs745937578 Chr.4: 170067368-170067368 on Build GRCh38
rs762424880 Chr.4: 170067412-170067412 on Build GRCh38
rs766009389 Chr.4: 170067426-170067426 on Build GRCh38
rs201801559 Chr.4: 170067302-170067302 on Build GRCh38

View Full Product Details