Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 119147348-119147619 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf30
Entrez Gene ID
152405
Gene Name
chromosome 3 open reading frame 30
Gene Aliases
TSCPA
UniGene
Hs.730099
Gene Chromosome Location
Chr.3: 119146150-119151455 on Build GRCh38
Transcripts
NM_152539.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3727345 c.1285A>T p.T429S Chr.3: 119147474-119147474 on Build GRCh38
5714192 c.1369G>A p.E457K Chr.3: 119147558-119147558 on Build GRCh38
4522519 c.1166G>A p.R389K Chr.3: 119147355-119147355 on Build GRCh38
7417092 c.1277C>T p.A426V Chr.3: 119147466-119147466 on Build GRCh38
6616013 c.1319T>G p.F440C Chr.3: 119147508-119147508 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145741636 Chr.3: 119147485-119147485 on Build GRCh38
rs986730093 Chr.3: 119147519-119147519 on Build GRCh38
rs754169437 Chr.3: 119147498-119147498 on Build GRCh38
rs770809358 Chr.3: 119147554-119147555 on Build GRCh38
rs56317615 Chr.3: 119147529-119147529 on Build GRCh38
rs1032241601 Chr.3: 119147385-119147385 on Build GRCh38
rs754105345 Chr.3: 119147544-119147544 on Build GRCh38
rs749217476 Chr.3: 119147512-119147512 on Build GRCh38
rs867439248 Chr.3: 119147518-119147518 on Build GRCh38
rs372941303 Chr.3: 119147375-119147375 on Build GRCh38
rs751466154 Chr.3: 119147548-119147548 on Build GRCh38
rs746924394 Chr.3: 119147424-119147424 on Build GRCh38
rs1031986761 Chr.3: 119147538-119147538 on Build GRCh38
rs765275130 Chr.3: 119147559-119147559 on Build GRCh38
rs769773425 Chr.3: 119147559-119147559 on Build GRCh38
rs199919487 Chr.3: 119147558-119147558 on Build GRCh38
rs754599571 Chr.3: 119147380-119147380 on Build GRCh38
rs759263930 Chr.3: 119147557-119147558 on Build GRCh38
rs1009309180 Chr.3: 119147581-119147581 on Build GRCh38
rs750474449 Chr.3: 119147566-119147566 on Build GRCh38
rs774149216 Chr.3: 119147555-119147556 on Build GRCh38
rs140682900 Chr.3: 119147412-119147412 on Build GRCh38
rs967414905 Chr.3: 119147459-119147459 on Build GRCh38
rs200587463 Chr.3: 119147543-119147543 on Build GRCh38
rs758995355 Chr.3: 119147567-119147568 on Build GRCh38
rs113445078 Chr.3: 119147456-119147456 on Build GRCh38
rs761106298 Chr.3: 119147477-119147477 on Build GRCh38
rs145826966 Chr.3: 119147572-119147572 on Build GRCh38
rs923407321 Chr.3: 119147506-119147506 on Build GRCh38
rs767296116 Chr.3: 119147558-119147558 on Build GRCh38
rs753437198 Chr.3: 119147377-119147377 on Build GRCh38
rs776777618 Chr.3: 119147558-119147559 on Build GRCh38
rs772753798 Chr.3: 119147502-119147503 on Build GRCh38
rs201169823 Chr.3: 119147555-119147555 on Build GRCh38
rs781321544 Chr.3: 119147554-119147554 on Build GRCh38
rs768972625 Chr.3: 119147442-119147442 on Build GRCh38
rs933531892 Chr.3: 119147448-119147448 on Build GRCh38
rs531656587 Chr.3: 119147565-119147565 on Build GRCh38
rs773725930 Chr.3: 119147476-119147476 on Build GRCh38
rs909223153 Chr.3: 119147388-119147388 on Build GRCh38
rs762537004 Chr.3: 119147596-119147596 on Build GRCh38
rs773750858 Chr.3: 119147436-119147436 on Build GRCh38
rs761943967 Chr.3: 119147464-119147464 on Build GRCh38
rs772138717 Chr.3: 119147566-119147566 on Build GRCh38
rs774785744 Chr.3: 119147447-119147447 on Build GRCh38
rs777180745 Chr.3: 119147545-119147545 on Build GRCh38
rs757241103 Chr.3: 119147552-119147552 on Build GRCh38
rs781158124 Chr.3: 119147438-119147438 on Build GRCh38
rs991476716 Chr.3: 119147374-119147374 on Build GRCh38
rs201518969 Chr.3: 119147474-119147474 on Build GRCh38
rs199894612 Chr.3: 119147564-119147564 on Build GRCh38
rs778574881 Chr.3: 119147392-119147392 on Build GRCh38
rs752354946 Chr.3: 119147583-119147583 on Build GRCh38
rs759958869 Chr.3: 119147504-119147504 on Build GRCh38
rs889335089 Chr.3: 119147486-119147486 on Build GRCh38
rs1036883495 Chr.3: 119147410-119147410 on Build GRCh38
rs199978595 Chr.3: 119147582-119147582 on Build GRCh38
rs777708183 Chr.3: 119147421-119147421 on Build GRCh38
rs551734199 Chr.3: 119147395-119147395 on Build GRCh38
rs78387691 Chr.3: 119147567-119147567 on Build GRCh38
rs549882362 Chr.3: 119147570-119147570 on Build GRCh38
rs149798586 Chr.3: 119147441-119147441 on Build GRCh38
rs368952787 Chr.3: 119147491-119147491 on Build GRCh38

View Full Product Details