Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 15442660-15442887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C1QTNF7
Entrez Gene ID
114905
Gene Name
C1q and tumor necrosis factor related protein 7
Gene Aliases
CTRP7, ZACRP7
UniGene
Hs.153714
Gene Chromosome Location
Chr.4: 15339818-15446169 on Build GRCh38
Transcripts
NM_001135170.1 NM_001135171.1 NM_031911.4 XM_011513772.1 XM_011513773.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
732352 c.809G>C p.G270A Chr.4: 15442738-15442738 on Build GRCh38
1618494 c.839T>C p.L280P Chr.4: 15442768-15442768 on Build GRCh38
3128354 c.803T>C p.F268S Chr.4: 15442732-15442732 on Build GRCh38
447512 c.774C>G p.F258L Chr.4: 15442703-15442703 on Build GRCh38
3128355 c.845C>T p.S282F Chr.4: 15442774-15442774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768048024 Chr.4: 15442803-15442803 on Build GRCh38
rs370436701 Chr.4: 15442842-15442842 on Build GRCh38
rs755297630 Chr.4: 15442764-15442764 on Build GRCh38
rs769646896 Chr.4: 15442770-15442770 on Build GRCh38
rs958438283 Chr.4: 15442812-15442812 on Build GRCh38
rs776483061 Chr.4: 15442752-15442752 on Build GRCh38
rs756544517 Chr.4: 15442832-15442832 on Build GRCh38
rs771855294 Chr.4: 15442745-15442745 on Build GRCh38
rs777288466 Chr.4: 15442747-15442747 on Build GRCh38
rs144053539 Chr.4: 15442736-15442736 on Build GRCh38
rs150973007 Chr.4: 15442841-15442841 on Build GRCh38
rs756593841 Chr.4: 15442722-15442722 on Build GRCh38
rs146900806 Chr.4: 15442737-15442737 on Build GRCh38
rs746533641 Chr.4: 15442749-15442749 on Build GRCh38
rs139276266 Chr.4: 15442776-15442776 on Build GRCh38
rs371568989 Chr.4: 15442719-15442719 on Build GRCh38
rs777329984 Chr.4: 15442844-15442844 on Build GRCh38
rs751031331 Chr.4: 15442831-15442831 on Build GRCh38
rs761842145 Chr.4: 15442800-15442800 on Build GRCh38
rs757730685 Chr.4: 15442689-15442689 on Build GRCh38
rs781578486 Chr.4: 15442706-15442706 on Build GRCh38
rs763098941 Chr.4: 15442774-15442774 on Build GRCh38
rs372133082 Chr.4: 15442758-15442758 on Build GRCh38
rs983788464 Chr.4: 15442765-15442765 on Build GRCh38
rs144156971 Chr.4: 15442751-15442751 on Build GRCh38
rs140830460 Chr.4: 15442862-15442862 on Build GRCh38
rs775273151 Chr.4: 15442773-15442773 on Build GRCh38
rs751621752 Chr.4: 15442797-15442797 on Build GRCh38
rs780499941 Chr.4: 15442838-15442838 on Build GRCh38

View Full Product Details