Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 113360470-113360715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BSPRY
Entrez Gene ID
54836
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
UniGene
Hs.614517
Gene Chromosome Location
Chr.9: 113349532-113371233 on Build GRCh38
Transcripts
NM_001317943.1 NM_001317944.1 NM_017688.2 XM_006717149.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1104220 c.415G>C p.A139P Chr.9: 113360621-113360621 on Build GRCh38
3653203 c.454G>A p.E152K Chr.9: 113360660-113360660 on Build GRCh38
4960928 c.450G>A p.W150* Chr.9: 113360656-113360656 on Build GRCh38
455183 c.468A>G p.K156K Chr.9: 113360674-113360674 on Build GRCh38
1459409 c.458C>T p.A153V Chr.9: 113360664-113360664 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767557160 Chr.9: 113360675-113360675 on Build GRCh38
rs777484954 Chr.9: 113360616-113360616 on Build GRCh38
rs777695975 Chr.9: 113360524-113360524 on Build GRCh38
rs762172231 Chr.9: 113360579-113360579 on Build GRCh38
rs771381539 Chr.9: 113360670-113360670 on Build GRCh38
rs570967595 Chr.9: 113360660-113360660 on Build GRCh38
rs755236113 Chr.9: 113360513-113360513 on Build GRCh38
rs375924305 Chr.9: 113360543-113360543 on Build GRCh38
rs773758644 Chr.9: 113360585-113360585 on Build GRCh38
rs142413897 Chr.9: 113360575-113360575 on Build GRCh38
rs766418761 Chr.9: 113360604-113360604 on Build GRCh38
rs778147226 Chr.9: 113360664-113360664 on Build GRCh38
rs757298690 Chr.9: 113360619-113360619 on Build GRCh38
rs376929788 Chr.9: 113360659-113360659 on Build GRCh38
rs751601032 Chr.9: 113360608-113360608 on Build GRCh38
rs757139288 Chr.9: 113360693-113360693 on Build GRCh38
rs757859006 Chr.9: 113360647-113360647 on Build GRCh38
rs372322975 Chr.9: 113360586-113360586 on Build GRCh38
rs770123180 Chr.9: 113360581-113360581 on Build GRCh38
rs773608048 Chr.9: 113360665-113360665 on Build GRCh38
rs753644953 Chr.9: 113360691-113360691 on Build GRCh38
rs770174612 Chr.9: 113360661-113360661 on Build GRCh38
rs771810350 Chr.9: 113360570-113360570 on Build GRCh38
rs767724502 Chr.9: 113360609-113360609 on Build GRCh38
rs750452809 Chr.9: 113360695-113360695 on Build GRCh38
rs752863534 Chr.9: 113360614-113360614 on Build GRCh38
rs766505291 Chr.9: 113360499-113360499 on Build GRCh38
rs745484136 Chr.9: 113360569-113360569 on Build GRCh38
rs752757 Chr.9: 113360674-113360674 on Build GRCh38
rs751760188 Chr.9: 113360511-113360511 on Build GRCh38
rs749516228 Chr.9: 113360544-113360544 on Build GRCh38
rs765206941 Chr.9: 113360694-113360694 on Build GRCh38
rs752492031 Chr.9: 113360523-113360523 on Build GRCh38
rs200736083 Chr.9: 113360687-113360687 on Build GRCh38
rs755929427 Chr.9: 113360615-113360615 on Build GRCh38
rs548730981 Chr.9: 113360688-113360688 on Build GRCh38
rs200697650 Chr.9: 113360576-113360576 on Build GRCh38
rs368339422 Chr.9: 113360564-113360564 on Build GRCh38
rs780270233 Chr.9: 113360539-113360539 on Build GRCh38
rs936491020 Chr.9: 113360534-113360534 on Build GRCh38
rs779023901 Chr.9: 113360631-113360631 on Build GRCh38
rs774857753 Chr.9: 113360672-113360672 on Build GRCh38
rs753802077 Chr.9: 113360618-113360618 on Build GRCh38
rs745322208 Chr.9: 113360640-113360640 on Build GRCh38
rs372300464 Chr.9: 113360522-113360522 on Build GRCh38

View Full Product Details