Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64949684-64949949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAJIN
Entrez Gene ID
283129
Gene Name
membrane anchored junction protein
Gene Aliases
C11orf85
UniGene
Hs.567793
Gene Chromosome Location
Chr.11: 64938230-64972104 on Build GRCh38
Transcripts
NM_001037225.2 NM_001300803.1 NM_001318808.1 XM_005273918.2 XM_006718502.2 XM_006718503.3 XM_006718504.2 XM_006718505.2 XM_011544940.2 XM_011544941.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
159464 c.285C>G p.I95M Chr.11: 64949807-64949807 on Build GRCh38
5499103 c.303delT p.F101fs*7 Chr.11: 64949789-64949789 on Build GRCh38
6023212 c.29C>T p.P10L Chr.11: 64949745-64949745 on Build GRCh38
6023211 c.347C>T p.P116L Chr.11: 64949745-64949745 on Build GRCh38
7104871 c.261C>G p.F87L Chr.11: 64949831-64949831 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971680012 Chr.11: 64949785-64949785 on Build GRCh38
rs772725014 Chr.11: 64949893-64949893 on Build GRCh38
rs748065665 Chr.11: 64949874-64949874 on Build GRCh38
rs771930121 Chr.11: 64949876-64949876 on Build GRCh38
rs761208556 Chr.11: 64949754-64949754 on Build GRCh38
rs753227681 Chr.11: 64949802-64949802 on Build GRCh38
rs774060709 Chr.11: 64949869-64949869 on Build GRCh38
rs765649508 Chr.11: 64949916-64949916 on Build GRCh38
rs528152528 Chr.11: 64949921-64949921 on Build GRCh38
rs775199136 Chr.11: 64949745-64949745 on Build GRCh38
rs958726737 Chr.11: 64949868-64949868 on Build GRCh38
rs559530258 Chr.11: 64949863-64949863 on Build GRCh38
rs374686374 Chr.11: 64949780-64949780 on Build GRCh38
rs772161531 Chr.11: 64949718-64949718 on Build GRCh38
rs901865679 Chr.11: 64949852-64949852 on Build GRCh38
rs192107829 Chr.11: 64949899-64949899 on Build GRCh38
rs776905673 Chr.11: 64949799-64949799 on Build GRCh38
rs767006767 Chr.11: 64949828-64949833 on Build GRCh38
rs762748424 Chr.11: 64949747-64949747 on Build GRCh38
rs893284156 Chr.11: 64949754-64949754 on Build GRCh38
rs777869823 Chr.11: 64949821-64949821 on Build GRCh38
rs781217503 Chr.11: 64949892-64949892 on Build GRCh38
rs765739578 Chr.11: 64949800-64949800 on Build GRCh38
rs755392453 Chr.11: 64949796-64949796 on Build GRCh38
rs781377867 Chr.11: 64949831-64949831 on Build GRCh38
rs758852369 Chr.11: 64949812-64949812 on Build GRCh38
rs759313336 Chr.11: 64949903-64949903 on Build GRCh38
rs747063337 Chr.11: 64949824-64949824 on Build GRCh38
rs868451871 Chr.11: 64949801-64949801 on Build GRCh38
rs143943051 Chr.11: 64949787-64949787 on Build GRCh38
rs373389251 Chr.11: 64949829-64949829 on Build GRCh38
rs768496335 Chr.11: 64949753-64949753 on Build GRCh38
rs777765112 Chr.11: 64949722-64949722 on Build GRCh38
rs200922605 Chr.11: 64949762-64949762 on Build GRCh38
rs767027510 Chr.11: 64949775-64949775 on Build GRCh38
rs907804614 Chr.11: 64949752-64949752 on Build GRCh38
rs377563243 Chr.11: 64949901-64949901 on Build GRCh38
rs369026602 Chr.11: 64949786-64949786 on Build GRCh38
rs745978635 Chr.11: 64949898-64949898 on Build GRCh38
rs1020933641 Chr.11: 64949784-64949784 on Build GRCh38
rs749920457 Chr.11: 64949778-64949778 on Build GRCh38
rs932045429 Chr.11: 64949878-64949878 on Build GRCh38
rs140011810 Chr.11: 64949704-64949704 on Build GRCh38
rs150526932 Chr.11: 64949742-64949742 on Build GRCh38
rs747205692 Chr.11: 64949733-64949733 on Build GRCh38
rs138283481 Chr.11: 64949756-64949756 on Build GRCh38
rs746154851 Chr.11: 64949861-64949861 on Build GRCh38

View Full Product Details