Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 92420961-92421232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLLU1OS
Entrez Gene ID
574016
Gene Name
chronic lymphocytic leukemia up-regulated 1 opposite strand
Gene Aliases
-
UniGene
Hs.551199
Gene Chromosome Location
Chr.12: 92420086-92428449 on Build GRCh38
Transcripts
NM_001025232.1 XM_017019673.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1323017 c.154A>T p.T52S Chr.12: 92421162-92421162 on Build GRCh38
1201418 c.303G>T p.M101I Chr.12: 92421013-92421013 on Build GRCh38
7183491 c.237G>C p.V79V Chr.12: 92421079-92421079 on Build GRCh38
695444 c.274G>A p.D92N Chr.12: 92421042-92421042 on Build GRCh38
4045393 c.151C>T p.L51L Chr.12: 92421165-92421165 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs181565725 Chr.12: 92421029-92421029 on Build GRCh38
rs769772491 Chr.12: 92421092-92421092 on Build GRCh38
rs768858889 Chr.12: 92421082-92421082 on Build GRCh38
rs149552124 Chr.12: 92421198-92421198 on Build GRCh38
rs746392706 Chr.12: 92421043-92421043 on Build GRCh38
rs914719238 Chr.12: 92421013-92421013 on Build GRCh38
rs962744126 Chr.12: 92421153-92421153 on Build GRCh38
rs370946717 Chr.12: 92420985-92420985 on Build GRCh38
rs1012778883 Chr.12: 92421048-92421048 on Build GRCh38
rs570396072 Chr.12: 92421163-92421163 on Build GRCh38
rs761897046 Chr.12: 92421087-92421087 on Build GRCh38
rs1000249161 Chr.12: 92421026-92421026 on Build GRCh38
rs74994222 Chr.12: 92421025-92421025 on Build GRCh38
rs144075926 Chr.12: 92421094-92421094 on Build GRCh38
rs781411843 Chr.12: 92421199-92421199 on Build GRCh38
rs770733032 Chr.12: 92421187-92421187 on Build GRCh38
rs762539325 Chr.12: 92421014-92421014 on Build GRCh38
rs147336226 Chr.12: 92421095-92421095 on Build GRCh38
rs777610728 Chr.12: 92421010-92421010 on Build GRCh38
rs1016053874 Chr.12: 92421164-92421164 on Build GRCh38
rs767611715 Chr.12: 92421111-92421111 on Build GRCh38
rs766330326 Chr.12: 92420984-92420984 on Build GRCh38
rs780603400 Chr.12: 92421182-92421182 on Build GRCh38
rs763781287 Chr.12: 92421149-92421149 on Build GRCh38
rs374170068 Chr.12: 92421191-92421191 on Build GRCh38
rs751468307 Chr.12: 92420992-92420992 on Build GRCh38
rs753435803 Chr.12: 92421154-92421154 on Build GRCh38
rs747409753 Chr.12: 92421076-92421076 on Build GRCh38
rs780370740 Chr.12: 92421071-92421071 on Build GRCh38
rs779408746 Chr.12: 92421041-92421041 on Build GRCh38
rs767455890 Chr.12: 92420999-92420999 on Build GRCh38
rs894353074 Chr.12: 92421030-92421030 on Build GRCh38
rs752511412 Chr.12: 92421141-92421141 on Build GRCh38
rs760615740 Chr.12: 92421145-92421145 on Build GRCh38
rs1026206999 Chr.12: 92421203-92421203 on Build GRCh38
rs759312047 Chr.12: 92420998-92420998 on Build GRCh38
rs200195927 Chr.12: 92421197-92421197 on Build GRCh38
rs758867349 Chr.12: 92421175-92421175 on Build GRCh38
rs7312566 Chr.12: 92421055-92421055 on Build GRCh38
rs776773541 Chr.12: 92421086-92421086 on Build GRCh38
rs756750757 Chr.12: 92421158-92421158 on Build GRCh38
rs867451070 Chr.12: 92421093-92421093 on Build GRCh38
rs755807168 Chr.12: 92421009-92421009 on Build GRCh38
rs201653871 Chr.12: 92421003-92421003 on Build GRCh38

View Full Product Details