Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 45030005-45030252 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAKD
Entrez Gene ID
79877
Gene Name
dephospho-CoA kinase domain containing
Gene Aliases
-
UniGene
Hs.463148
Gene Chromosome Location
Chr.17: 45023338-45061129 on Build GRCh38
Transcripts
NM_001128631.2 NM_001288654.2 NM_001288655.1 NM_001321326.1 NM_024819.6 XM_011525262.2 XM_017025102.1 XM_017025104.1 XM_017025105.1 XM_017025106.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6497795 c.345C>T p.P115P Chr.17: 45030151-45030151 on Build GRCh38
7414040 c.377A>C p.Y126S Chr.17: 45030119-45030119 on Build GRCh38
706107 c.326A>G p.Y109C Chr.17: 45030170-45030170 on Build GRCh38
3191819 c.327C>T p.Y109Y Chr.17: 45030169-45030169 on Build GRCh38
3191818 c.386A>C p.H129P Chr.17: 45030110-45030110 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs550847925 Chr.17: 45030052-45030052 on Build GRCh38
rs758715593 Chr.17: 45030177-45030177 on Build GRCh38
rs749937712 Chr.17: 45030169-45030169 on Build GRCh38
rs768375559 Chr.17: 45030228-45030228 on Build GRCh38
rs73311379 Chr.17: 45030221-45030221 on Build GRCh38
rs374665325 Chr.17: 45030198-45030198 on Build GRCh38
rs775423308 Chr.17: 45030142-45030142 on Build GRCh38
rs867161523 Chr.17: 45030133-45030133 on Build GRCh38
rs987207831 Chr.17: 45030130-45030130 on Build GRCh38
rs1024118921 Chr.17: 45030102-45030102 on Build GRCh38
rs762661456 Chr.17: 45030147-45030147 on Build GRCh38
rs767094501 Chr.17: 45030073-45030073 on Build GRCh38
rs140561456 Chr.17: 45030033-45030033 on Build GRCh38
rs755803493 Chr.17: 45030077-45030077 on Build GRCh38
rs201909000 Chr.17: 45030075-45030075 on Build GRCh38
rs757743341 Chr.17: 45030188-45030188 on Build GRCh38
rs751475319 Chr.17: 45030056-45030056 on Build GRCh38
rs753468881 Chr.17: 45030175-45030175 on Build GRCh38
rs755858662 Chr.17: 45030170-45030170 on Build GRCh38
rs144472327 Chr.17: 45030174-45030174 on Build GRCh38
rs769845706 Chr.17: 45030205-45030205 on Build GRCh38
rs750139691 Chr.17: 45030074-45030074 on Build GRCh38
rs150982403 Chr.17: 45030105-45030105 on Build GRCh38
rs767182459 Chr.17: 45030168-45030168 on Build GRCh38
rs763891925 Chr.17: 45030150-45030150 on Build GRCh38
rs778164914 Chr.17: 45030180-45030180 on Build GRCh38
rs72255860 Chr.17: 45030082-45030083 on Build GRCh38
rs774264492 Chr.17: 45030229-45030229 on Build GRCh38
rs774461028 Chr.17: 45030154-45030154 on Build GRCh38
rs748453279 Chr.17: 45030096-45030096 on Build GRCh38
rs533128602 Chr.17: 45030110-45030110 on Build GRCh38
rs199748913 Chr.17: 45030106-45030106 on Build GRCh38
rs761810256 Chr.17: 45030159-45030159 on Build GRCh38
rs763006768 Chr.17: 45030044-45030044 on Build GRCh38
rs761898110 Chr.17: 45030231-45030231 on Build GRCh38
rs749485783 Chr.17: 45030222-45030222 on Build GRCh38
rs979860782 Chr.17: 45030166-45030166 on Build GRCh38
rs769897043 Chr.17: 45030135-45030135 on Build GRCh38
rs780116609 Chr.17: 45030068-45030068 on Build GRCh38
rs201327450 Chr.17: 45030191-45030191 on Build GRCh38
rs779390345 Chr.17: 45030082-45030082 on Build GRCh38
rs886371524 Chr.17: 45030034-45030034 on Build GRCh38
rs142207624 Chr.17: 45030109-45030109 on Build GRCh38
rs758719106 Chr.17: 45030097-45030097 on Build GRCh38
rs768564793 Chr.17: 45030093-45030094 on Build GRCh38
rs142583348 Chr.17: 45030103-45030103 on Build GRCh38

View Full Product Details