Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67185297-67185522 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXOC3L1
Entrez Gene ID
283849
Gene Name
exocyst complex component 3 like 1
Gene Aliases
EXOC3L
UniGene
Hs.647356
Gene Chromosome Location
Chr.16: 67184379-67190204 on Build GRCh38
Transcripts
NM_178516.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
32711 c.1541C>A p.A514D Chr.16: 67185446-67185446 on Build GRCh38
6657114 c.1543T>C p.L515L Chr.16: 67185444-67185444 on Build GRCh38
1749694 c.1578G>A p.Q526Q Chr.16: 67185409-67185409 on Build GRCh38
138080 c.1618G>A p.E540K Chr.16: 67185369-67185369 on Build GRCh38
3712061 c.1580G>A p.R527K Chr.16: 67185407-67185407 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs914654967 Chr.16: 67185492-67185492 on Build GRCh38
rs1046893250 Chr.16: 67185343-67185343 on Build GRCh38
rs756423331 Chr.16: 67185476-67185476 on Build GRCh38
rs765261425 Chr.16: 67185459-67185459 on Build GRCh38
rs371403303 Chr.16: 67185353-67185353 on Build GRCh38
rs554282272 Chr.16: 67185472-67185472 on Build GRCh38
rs1008563482 Chr.16: 67185395-67185395 on Build GRCh38
rs774586129 Chr.16: 67185437-67185437 on Build GRCh38
rs575513337 Chr.16: 67185460-67185460 on Build GRCh38
rs768672619 Chr.16: 67185434-67185434 on Build GRCh38
rs138861636 Chr.16: 67185461-67185461 on Build GRCh38
rs763059087 Chr.16: 67185441-67185441 on Build GRCh38
rs752062351 Chr.16: 67185425-67185425 on Build GRCh38
rs766121776 Chr.16: 67185325-67185325 on Build GRCh38
rs776547426 Chr.16: 67185449-67185449 on Build GRCh38
rs138391550 Chr.16: 67185377-67185377 on Build GRCh38
rs151131184 Chr.16: 67185482-67185482 on Build GRCh38
rs768754815 Chr.16: 67185489-67185489 on Build GRCh38
rs142067275 Chr.16: 67185451-67185451 on Build GRCh38
rs924793766 Chr.16: 67185469-67185469 on Build GRCh38
rs746614088 Chr.16: 67185396-67185396 on Build GRCh38
rs760484018 Chr.16: 67185321-67185321 on Build GRCh38
rs373250658 Chr.16: 67185316-67185316 on Build GRCh38
rs749432554 Chr.16: 67185429-67185429 on Build GRCh38
rs774365905 Chr.16: 67185495-67185495 on Build GRCh38
rs375469444 Chr.16: 67185406-67185406 on Build GRCh38
rs751264822 Chr.16: 67185465-67185465 on Build GRCh38
rs746200743 Chr.16: 67185413-67185413 on Build GRCh38
rs142958191 Chr.16: 67185446-67185446 on Build GRCh38
rs757731618 Chr.16: 67185398-67185398 on Build GRCh38
rs770395508 Chr.16: 67185414-67185414 on Build GRCh38
rs141493416 Chr.16: 67185466-67185466 on Build GRCh38
rs753723469 Chr.16: 67185330-67185330 on Build GRCh38
rs763660963 Chr.16: 67185462-67185462 on Build GRCh38
rs766175324 Chr.16: 67185448-67185448 on Build GRCh38
rs757817997 Chr.16: 67185369-67185369 on Build GRCh38
rs553497676 Chr.16: 67185349-67185349 on Build GRCh38
rs1021617513 Chr.16: 67185447-67185447 on Build GRCh38
rs996889956 Chr.16: 67185432-67185432 on Build GRCh38
rs754887106 Chr.16: 67185335-67185335 on Build GRCh38
rs766534906 Chr.16: 67185470-67185471 on Build GRCh38

View Full Product Details