Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 6970871-6971145 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMG1
Entrez Gene ID
10436
Gene Name
EMG1, N1-specific pseudouridine methyltransferase
Gene Aliases
C2F, Grcc2f, NEP1
UniGene
Hs.558447
Gene Chromosome Location
Chr.12: 6970781-6997428 on Build GRCh38
Transcripts
NM_001320049.1 NM_006331.7 NR_135131.1

SNP Detail

SNP ID Chromosome Location
rs36061201 Chr.12: 6970961-6970961 on Build GRCh38
rs782249279 Chr.12: 6970965-6970965 on Build GRCh38
rs782778398 Chr.12: 6970943-6970943 on Build GRCh38
rs782597296 Chr.12: 6970901-6970901 on Build GRCh38
rs781824161 Chr.12: 6971036-6971036 on Build GRCh38
rs782186328 Chr.12: 6971058-6971058 on Build GRCh38
rs782250795 Chr.12: 6971031-6971031 on Build GRCh38
rs782103478 Chr.12: 6971075-6971075 on Build GRCh38
rs782150637 Chr.12: 6970982-6970982 on Build GRCh38
rs782686642 Chr.12: 6971046-6971046 on Build GRCh38
rs782628939 Chr.12: 6970960-6970960 on Build GRCh38
rs782752845 Chr.12: 6970896-6970896 on Build GRCh38
rs782611143 Chr.12: 6971085-6971085 on Build GRCh38
rs11064480 Chr.12: 6971024-6971024 on Build GRCh38
rs782743040 Chr.12: 6971005-6971005 on Build GRCh38
rs782175380 Chr.12: 6970966-6970966 on Build GRCh38
rs781851789 Chr.12: 6971012-6971012 on Build GRCh38
rs1003756868 Chr.12: 6971006-6971006 on Build GRCh38
rs371837043 Chr.12: 6970908-6970908 on Build GRCh38
rs781966739 Chr.12: 6971071-6971071 on Build GRCh38
rs782780843 Chr.12: 6970904-6970904 on Build GRCh38
rs60117710 Chr.12: 6971047-6971048 on Build GRCh38
rs782550075 Chr.12: 6970947-6970947 on Build GRCh38
rs782016119 Chr.12: 6971082-6971082 on Build GRCh38
rs782075048 Chr.12: 6971003-6971003 on Build GRCh38
rs782546340 Chr.12: 6970964-6970964 on Build GRCh38
rs782541564 Chr.12: 6970900-6970900 on Build GRCh38
rs782083207 Chr.12: 6971102-6971102 on Build GRCh38
rs782429979 Chr.12: 6970929-6970929 on Build GRCh38
rs781892156 Chr.12: 6970976-6970977 on Build GRCh38
rs782524772 Chr.12: 6971019-6971020 on Build GRCh38
rs782055621 Chr.12: 6970947-6970948 on Build GRCh38
rs782500790 Chr.12: 6971037-6971037 on Build GRCh38
rs782711576 Chr.12: 6970940-6970940 on Build GRCh38
rs782604752 Chr.12: 6970970-6970970 on Build GRCh38
rs149578211 Chr.12: 6970938-6970938 on Build GRCh38
rs781893222 Chr.12: 6970944-6970944 on Build GRCh38
rs781925110 Chr.12: 6970996-6970996 on Build GRCh38
rs200041551 Chr.12: 6971047-6971047 on Build GRCh38
rs142280686 Chr.12: 6971120-6971120 on Build GRCh38
rs782233712 Chr.12: 6970975-6970975 on Build GRCh38
rs982421993 Chr.12: 6971073-6971073 on Build GRCh38
rs782273494 Chr.12: 6970921-6970921 on Build GRCh38
rs782324364 Chr.12: 6970969-6970969 on Build GRCh38
rs372361920 Chr.12: 6970955-6970955 on Build GRCh38
rs782479150 Chr.12: 6970954-6970954 on Build GRCh38
rs375042715 Chr.12: 6971077-6971077 on Build GRCh38
rs782633987 Chr.12: 6971039-6971039 on Build GRCh38
rs782266941 Chr.12: 6971044-6971044 on Build GRCh38
rs782722332 Chr.12: 6970950-6970950 on Build GRCh38
rs781804386 Chr.12: 6971096-6971096 on Build GRCh38
rs782222037 Chr.12: 6970902-6970902 on Build GRCh38
rs781814528 Chr.12: 6970898-6970898 on Build GRCh38
rs952438354 Chr.12: 6971032-6971032 on Build GRCh38
rs369617092 Chr.12: 6970978-6970978 on Build GRCh38
rs782695884 Chr.12: 6971088-6971088 on Build GRCh38
rs368907259 Chr.12: 6971101-6971101 on Build GRCh38
rs376055535 Chr.12: 6970936-6970936 on Build GRCh38
rs906784380 Chr.12: 6970986-6970986 on Build GRCh38
rs782126924 Chr.12: 6971014-6971014 on Build GRCh38
rs782438220 Chr.12: 6970984-6970984 on Build GRCh38
rs782414000 Chr.12: 6971045-6971045 on Build GRCh38
rs781917483 Chr.12: 6970935-6970935 on Build GRCh38
rs782806796 Chr.12: 6970967-6970968 on Build GRCh38
rs376797135 Chr.12: 6971092-6971092 on Build GRCh38
rs782118451 Chr.12: 6970942-6970942 on Build GRCh38
rs17857448 Chr.12: 6971049-6971049 on Build GRCh38
rs3168594 Chr.12: 6971027-6971027 on Build GRCh38

View Full Product Details