Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 100803406-100803635 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPHB4
Entrez Gene ID
2050
Gene Name
EPH receptor B4
Gene Aliases
HTK, MYK1, TYRO11
UniGene
Hs.437008
Gene Chromosome Location
Chr.7: 100802565-100827521 on Build GRCh38
Transcripts
NM_004444.4 XM_017011816.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4833018 c.2862G>A p.L954L Chr.7: 100803563-100803563 on Build GRCh38
3876458 c.2953C>A p.P985T Chr.7: 100803472-100803472 on Build GRCh38
4982103 c.2949G>A p.P983P Chr.7: 100803476-100803476 on Build GRCh38
1083128 c.2949_2950insACCG p.A984fs*>6 Chr.7: 100803475-100803476 on Build GRCh38
5500433 c.2882T>C p.I961T Chr.7: 100803543-100803543 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773050327 Chr.7: 100803495-100803495 on Build GRCh38
rs564273618 Chr.7: 100803503-100803503 on Build GRCh38
rs767280149 Chr.7: 100803532-100803532 on Build GRCh38
rs199872533 Chr.7: 100803430-100803430 on Build GRCh38
rs759204782 Chr.7: 100803515-100803515 on Build GRCh38
rs1048885957 Chr.7: 100803501-100803501 on Build GRCh38
rs998394553 Chr.7: 100803587-100803587 on Build GRCh38
rs140842916 Chr.7: 100803471-100803471 on Build GRCh38
rs777665533 Chr.7: 100803614-100803614 on Build GRCh38
rs889742064 Chr.7: 100803522-100803522 on Build GRCh38
rs56086910 Chr.7: 100803470-100803470 on Build GRCh38
rs758969373 Chr.7: 100803586-100803586 on Build GRCh38
rs769456757 Chr.7: 100803610-100803610 on Build GRCh38
rs781304760 Chr.7: 100803603-100803603 on Build GRCh38
rs766227428 Chr.7: 100803584-100803584 on Build GRCh38
rs768926072 Chr.7: 100803477-100803477 on Build GRCh38
rs937269398 Chr.7: 100803440-100803440 on Build GRCh38
rs751237774 Chr.7: 100803434-100803434 on Build GRCh38
rs754741567 Chr.7: 100803443-100803443 on Build GRCh38
rs1007131202 Chr.7: 100803517-100803517 on Build GRCh38
rs769735270 Chr.7: 100803485-100803485 on Build GRCh38
rs112803997 Chr.7: 100803476-100803476 on Build GRCh38
rs754186641 Chr.7: 100803560-100803560 on Build GRCh38
rs770536938 Chr.7: 100803505-100803505 on Build GRCh38
rs776048181 Chr.7: 100803494-100803494 on Build GRCh38
rs754162681 Chr.7: 100803460-100803460 on Build GRCh38
rs879117903 Chr.7: 100803445-100803445 on Build GRCh38
rs751653870 Chr.7: 100803593-100803593 on Build GRCh38
rs747900634 Chr.7: 100803479-100803479 on Build GRCh38
rs192294324 Chr.7: 100803445-100803445 on Build GRCh38
rs534188389 Chr.7: 100803513-100803513 on Build GRCh38
rs747930613 Chr.7: 100803607-100803607 on Build GRCh38
rs755125949 Chr.7: 100803600-100803600 on Build GRCh38
rs750862933 Chr.7: 100803580-100803580 on Build GRCh38
rs780095931 Chr.7: 100803588-100803588 on Build GRCh38
rs545342926 Chr.7: 100803456-100803456 on Build GRCh38
rs765896379 Chr.7: 100803495-100803495 on Build GRCh38
rs1046460658 Chr.7: 100803478-100803478 on Build GRCh38
rs991387940 Chr.7: 100803441-100803441 on Build GRCh38
rs139196073 Chr.7: 100803561-100803561 on Build GRCh38
rs373446670 Chr.7: 100803579-100803579 on Build GRCh38

View Full Product Details