Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66940311-66940546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES2
Entrez Gene ID
8824
Gene Name
carboxylesterase 2
Gene Aliases
CE-2, CES2A1, PCE-2, iCE
UniGene
Hs.282975
Gene Chromosome Location
Chr.16: 66934444-66945096 on Build GRCh38
Transcripts
NM_003869.5 NM_198061.2 NR_036684.1 XM_011523421.2 XM_017023818.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
194317 c.808C>T p.R270C Chr.16: 66940495-66940495 on Build GRCh38
5851189 c.838G>T p.G280* Chr.16: 66940525-66940525 on Build GRCh38
1478959 c.724C>T p.R242C Chr.16: 66940330-66940330 on Build GRCh38
6273124 c.733G>T p.V245F Chr.16: 66940339-66940339 on Build GRCh38
213700 c.790G>A p.D264N Chr.16: 66940477-66940477 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750420342 Chr.16: 66940505-66940505 on Build GRCh38
rs755562282 Chr.16: 66940512-66940512 on Build GRCh38
rs985457932 Chr.16: 66940494-66940494 on Build GRCh38
rs767322191 Chr.16: 66940501-66940501 on Build GRCh38
rs565364625 Chr.16: 66940331-66940331 on Build GRCh38
rs377418721 Chr.16: 66940381-66940381 on Build GRCh38
rs749857196 Chr.16: 66940421-66940421 on Build GRCh38
rs759686749 Chr.16: 66940477-66940477 on Build GRCh38
rs111381920 Chr.16: 66940488-66940488 on Build GRCh38
rs370961425 Chr.16: 66940444-66940444 on Build GRCh38
rs8192924 Chr.16: 66940496-66940496 on Build GRCh38
rs964487090 Chr.16: 66940450-66940450 on Build GRCh38
rs754108533 Chr.16: 66940386-66940386 on Build GRCh38
rs112732183 Chr.16: 66940516-66940516 on Build GRCh38
rs769189373 Chr.16: 66940375-66940375 on Build GRCh38
rs537562880 Chr.16: 66940453-66940453 on Build GRCh38
rs760843453 Chr.16: 66940383-66940383 on Build GRCh38
rs772649560 Chr.16: 66940382-66940382 on Build GRCh38
rs765456699 Chr.16: 66940478-66940478 on Build GRCh38
rs532312387 Chr.16: 66940517-66940517 on Build GRCh38
rs779782688 Chr.16: 66940361-66940361 on Build GRCh38
rs755685960 Chr.16: 66940428-66940428 on Build GRCh38
rs769281134 Chr.16: 66940349-66940349 on Build GRCh38
rs201696544 Chr.16: 66940394-66940394 on Build GRCh38
rs990644871 Chr.16: 66940510-66940510 on Build GRCh38
rs754684353 Chr.16: 66940445-66940445 on Build GRCh38
rs957447788 Chr.16: 66940503-66940503 on Build GRCh38
rs913361346 Chr.16: 66940515-66940515 on Build GRCh38
rs757128242 Chr.16: 66940408-66940408 on Build GRCh38
rs762723950 Chr.16: 66940392-66940392 on Build GRCh38
rs767512037 Chr.16: 66940420-66940420 on Build GRCh38
rs1021357931 Chr.16: 66940433-66940433 on Build GRCh38
rs747038619 Chr.16: 66940459-66940459 on Build GRCh38
rs146178564 Chr.16: 66940458-66940458 on Build GRCh38
rs143299572 Chr.16: 66940452-66940452 on Build GRCh38
rs1011344581 Chr.16: 66940432-66940432 on Build GRCh38
rs200252180 Chr.16: 66940376-66940376 on Build GRCh38
rs776832259 Chr.16: 66940471-66940471 on Build GRCh38
rs550018149 Chr.16: 66940464-66940464 on Build GRCh38
rs761698722 Chr.16: 66940495-66940495 on Build GRCh38
rs189163624 Chr.16: 66940384-66940384 on Build GRCh38
rs201028689 Chr.16: 66940393-66940393 on Build GRCh38
rs749635127 Chr.16: 66940371-66940371 on Build GRCh38
rs561168882 Chr.16: 66940431-66940431 on Build GRCh38

View Full Product Details