Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 57575347-57575606 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARF4
Entrez Gene ID
378
Gene Name
ADP ribosylation factor 4
Gene Aliases
ARF2
UniGene
Hs.652183
Gene Chromosome Location
Chr.3: 57571363-57597488 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Gene Symbol
PDE12
Entrez Gene ID
201626
Gene Name
phosphodiesterase 12
Gene Aliases
2'-PDE, 2-PDE
UniGene
Hs.572993
Gene Chromosome Location
Chr.3: 57556247-57656480 on Build GRCh38
Transcripts
NM_001660.3 NM_001322176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7123406 c.419C>T p.T140I Chr.3: 57575585-57575585 on Build GRCh38
7114812 c.408C>A p.I136I Chr.3: 57575596-57575596 on Build GRCh38
6602849 c.456+2T>C p.? Chr.3: 57575546-57575546 on Build GRCh38
167927 c.445C>T p.R149C Chr.3: 57575559-57575559 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs772942523 Chr.3: 57575499-57575499 on Build GRCh38
rs747265204 Chr.3: 57575559-57575559 on Build GRCh38
rs371115651 Chr.3: 57575532-57575532 on Build GRCh38
rs760263492 Chr.3: 57575503-57575503 on Build GRCh38
rs982161495 Chr.3: 57575481-57575481 on Build GRCh38
rs752471163 Chr.3: 57575544-57575544 on Build GRCh38
rs989869645 Chr.3: 57575465-57575465 on Build GRCh38
rs746108580 Chr.3: 57575459-57575462 on Build GRCh38
rs567392195 Chr.3: 57575412-57575412 on Build GRCh38
rs777379437 Chr.3: 57575558-57575558 on Build GRCh38
rs536459530 Chr.3: 57575430-57575430 on Build GRCh38
rs758239795 Chr.3: 57575557-57575557 on Build GRCh38
rs969562807 Chr.3: 57575394-57575394 on Build GRCh38
rs958344481 Chr.3: 57575414-57575414 on Build GRCh38
rs754966249 Chr.3: 57575527-57575527 on Build GRCh38
rs765876714 Chr.3: 57575507-57575507 on Build GRCh38
rs148967593 Chr.3: 57575578-57575578 on Build GRCh38
rs980515110 Chr.3: 57575435-57575435 on Build GRCh38
rs781482985 Chr.3: 57575566-57575566 on Build GRCh38
rs528716213 Chr.3: 57575429-57575433 on Build GRCh38
rs753339388 Chr.3: 57575526-57575526 on Build GRCh38
rs556043399 Chr.3: 57575477-57575477 on Build GRCh38
rs916562333 Chr.3: 57575471-57575471 on Build GRCh38

View Full Product Details